Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01476820_m1
          See other GRAMD1A GE Assays ›
          Gene Symbol:
          Gramd1a
          Gene Name:
          GRAM domain containing 1A
          Gene Aliases:
          Eg1rvc, RGD1311022
          Chromosome Location:
          Chr.1: 89563982 - 89590410 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01476820_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gramd1a
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001014160.1 NP_001014182.1 17 - 18 2052 91
          XM_006228832.2 XP_006228894.1 20 - 21 2479 91
          XM_006228833.2 XP_006228895.1 19 - 20 2465 91
          XM_006228834.2 XP_006228896.1 19 - 20 2245 91
          XM_006228835.2 XP_006228897.1 18 - 19 2231 91
          GenBank mRNA AY171575.1 - 17 - 18 2052 91
          BC105896.1 - 18 - 19 2153 7380410 91

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100005186.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Gramd1a

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P321918 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 32739 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 148362 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 506201 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 3307 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 64585 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 413252 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 760317 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 819128 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 291197 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 189686 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 73659 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P321918 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P321918 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P321918 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P321918 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 2738 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 53220 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 45477 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10720933 - -
          Affymetrix Rat Gene 1.0 ST Array 10720921 - -
          Affymetrix Rat Gene 1.0 ST Array 10720919 - -
          Affymetrix Rat Gene 1.0 ST Array 10720930 - -
          Affymetrix Rat Gene 1.0 ST Array 10720936 - -
          Affymetrix Rat Gene 1.0 ST Array 10720920 - -
          Affymetrix Rat Gene 1.0 ST Array 10720932 - -
          Affymetrix Rat Gene 1.0 ST Array 10720924 - -
          Affymetrix Rat Gene 1.0 ST Array 10720935 - -
          Affymetrix Rat Gene 1.0 ST Array 10720925 - -
          Affymetrix Rat Gene 1.0 ST Array 10720931 - -
          Affymetrix Rat Gene 1.0 ST Array 10720926 - -
          Affymetrix Rat Gene 1.0 ST Array 10720928 - -
          Affymetrix Rat Gene 1.0 ST Array 10720923 - -
          Affymetrix Rat Gene 1.0 ST Array 10720922 - -
          Affymetrix Rat Gene 1.0 ST Array 10720927 - -
          Affymetrix Rat Gene 1.0 ST Array 10720934 - -
          Affymetrix Rat Gene 1.0 ST Array 10720929 - -
          Affymetrix Rat Gene 1.1 ST Array 10720918 - -
          Affymetrix Rat Clariom S Assay PSR0100041907.rn.1 TC0100005186.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100041906.rn.1 TC0100005186.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100041904.rn.1 TC0100005186.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100041905.rn.1 TC0100005186.rn.1 RTA TC
          Affymetrix Rat Gene 1.0 ST Array 10720918 - -
          Rat Genome U34 Set rc_AA819685_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20971361 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1356460 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-mrft7:80/100.66.76.145:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0