Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01477032_m1
          See other VASP GE Assays ›
          Gene Symbol:
          Vasp
          Gene Name:
          vasodilator-stimulated phosphoprotein
          Gene Aliases:
          -
          Chromosome Location:
          Chr.1: 80170608 - 80186458 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01477032_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Vasp
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001108475.2 NP_001101945.2 4 - 5 614 146
          XM_008758953.1 XP_008757175.1 4 - 5 1190 146
          GenBank mRNA BC168841.1 - 4 - 5 614 9030574 146

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100004913.rn.1

          Set Membership:

          Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Panther Classification:

          Molecular Function -

          scaffold/adaptor protein

          Gene Ontology Categories:

          Function(s) Process(es)

          neural tube closure
          axon guidance
          actin polymerization or depolymerization
          actin cytoskeleton organization
          positive regulation of actin filament polymerization
          protein homotetramerization
          actin binding
          profilin binding
          SH3 domain binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P198935 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 344348 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 390964 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 971408 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 829068 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 698397 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 685240 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 182893 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 25452 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 425487 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 486530 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 164033 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 44372 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P198935 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P198935 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P198935 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P198935 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 103741 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10719417 - -
          Affymetrix Rat Gene 1.0 ST Array 10719416 - -
          Affymetrix Rat Gene 1.0 ST Array 10719412 - -
          Affymetrix Rat Gene 1.0 ST Array 10719423 - -
          Affymetrix Rat Gene 1.0 ST Array 10719415 - -
          Affymetrix Rat Gene 1.0 ST Array 10719420 - -
          Affymetrix Rat Gene 1.0 ST Array 10719414 - -
          Affymetrix Rat Gene 1.0 ST Array 10719422 - -
          Affymetrix Rat Gene 1.0 ST Array 10719424 - -
          Affymetrix Rat Gene 1.0 ST Array 10719418 - -
          Affymetrix Rat Gene 1.0 ST Array 10719421 - -
          Affymetrix Rat Gene 1.0 ST Array 10719419 - -
          Affymetrix Rat Gene 1.0 ST Array 10719413 - -
          Affymetrix Rat Gene 1.1 ST Array 10719411 - -
          Rat Genome 230 2.0 Array 1372459_at - -
          Rat Genome 230 2.0 Array 1372459_PM_at - -
          Affymetrix HT Rat Focus Array 1372459_at - -
          Affymetrix HT Rat Focus Array 1372459_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0100038745.rn.1 TC0100004913.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100038746.rn.1 TC0100004913.rn.1 RTA TC
          Rat Expression Set 230 1372459_at - -
          Rat Expression Set 230 1372459_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10719411 - -
          Rat Genome U34 Set rc_AA956278_at - -
          Rat Genome U34 Set rc_AA956278_g_at - -
          Rat Genome U34 Set rc_AI177116_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21497458 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1370521 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-jwmpg:80/100.66.76.145:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0