Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01477185_m1
          See other SETD4 GE Assays ›
          Gene Symbol:
          Setd4
          Gene Name:
          SET domain containing 4
          Gene Aliases:
          Rda279
          Chromosome Location:
          Chr.11: 33781869 - 33803115 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01477185_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Setd4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001113747.1 NP_001107219.1 1020 67
          XM_006248028.2 XP_006248090.1 5318 67
          XM_008768559.1 XP_008766781.1 7536 67
          XM_008768560.1 XP_008766782.1 4314 67
          XM_008768561.1 XP_008766783.1 4047 67
          GenBank mRNA AF388528.1 - 1 - 2 393 67
          BC079469.1 - 856 67
          BC158669.1 - 1020 8370827 67

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1100001344.rn.1

          Set Membership:

          Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Panther Classification:

          Molecular Function -

          methyltransferase

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P1039678 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 500753 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 587825 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 212596 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 180441 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 829260 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 920871 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 20322 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 606775 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 498238 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 804098 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 395675 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 847180 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 74482 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 133906 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P1039678 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P1039678 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P1039678 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P1039678 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 10054 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 94955 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10753235 - -
          Affymetrix Rat Gene 1.0 ST Array 10753240 - -
          Affymetrix Rat Gene 1.0 ST Array 10753239 - -
          Affymetrix Rat Gene 1.0 ST Array 10753232 - -
          Affymetrix Rat Gene 1.0 ST Array 10753242 - -
          Affymetrix Rat Gene 1.0 ST Array 10753233 - -
          Affymetrix Rat Gene 1.0 ST Array 10753234 - -
          Affymetrix Rat Gene 1.0 ST Array 10753238 - -
          Affymetrix Rat Gene 1.0 ST Array 10753236 - -
          Affymetrix Rat Gene 1.0 ST Array 10753243 - -
          Affymetrix Rat Gene 1.0 ST Array 10753241 - -
          Affymetrix Rat Gene 1.1 ST Array 10753231 - -
          Rat Genome 230 2.0 Array 1370947_at - -
          Rat Genome 230 2.0 Array 1370947_PM_at - -
          Affymetrix HT Rat Focus Array 1370947_at - -
          Affymetrix HT Rat Focus Array 1370947_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1100008001.rn.1 TC1100001344.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1100008005.rn.1 TC1100001344.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1100007997.rn.1 TC1100001344.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1100008023.rn.1 TC1100001344.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1100008010.rn.1 TC1100001344.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1100007998.rn.1 TC1100001344.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1100008026.rn.1 TC1100001344.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1100008013.rn.1 TC1100001344.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1100008006.rn.1 TC1100001344.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1100008024.rn.1 TC1100001344.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1100008011.rn.1 TC1100001344.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1100008025.rn.1 TC1100001344.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1100008009.rn.1 TC1100001344.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1100008000.rn.1 TC1100001344.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1100007995.rn.1 TC1100001344.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1100008008.rn.1 TC1100001344.rn.1 RTA TC
          Rat Expression Set 230 1370947_PM_at - -
          Rat Expression Set 230 1370947_at - -
          Affymetrix Rat Gene 1.0 ST Array 10753231 - -
          Rat Genome U34 Set rc_AI228197_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21563553 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1358039 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline