Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Setd8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001246665.1 | NP_001233594.1 | 6 - 7 | 761 | 143 | |
XM_006249328.2 | XP_006249390.1 | 7 - 8 | 923 | 143 | ||
XM_006249329.2 | XP_006249391.1 | 7 - 8 | 893 | 143 | ||
XM_006249330.2 | XP_006249392.1 | 6 - 7 | 838 | 143 | ||
XM_008769264.1 | XP_008767486.1 | 6 - 7 | 845 | 143 | ||
GenBank mRNA | AB606422.1 | - | 6 - 7 | 761 | 143 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons |