Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01477432_m1
          See other TM2D3 GE Assays ›
          Gene Symbol:
          Tm2d3
          Gene Name:
          TM2 domain containing 3
          Gene Aliases:
          RGD1564929
          Chromosome Location:
          Chr.1: 126573474 - 126595778 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01477432_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tm2d3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001106267.1 NP_001099737.1 1153 73

          Back To Top

          More Information


          Set Membership:

          Amplicon length between 71 and 85 3' Most Recommended Assay Rattus norvegicus 1700 Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P057907 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 951469 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 880356 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 638944 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 459627 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 257972 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 711806 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 25275 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 125304 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 759403 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 61905 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P057907 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P057907 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P057907 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P1004470 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 54694 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10707785 - -
          Affymetrix Rat Gene 1.0 ST Array 10707783 - -
          Affymetrix Rat Gene 1.0 ST Array 10707780 - -
          Affymetrix Rat Gene 1.0 ST Array 10707786 - -
          Affymetrix Rat Gene 1.0 ST Array 10707781 - -
          Affymetrix Rat Gene 1.0 ST Array 10707787 - -
          Affymetrix Rat Gene 1.0 ST Array 10707784 - -
          Affymetrix Rat Gene 1.0 ST Array 10707782 - -
          Affymetrix Rat Gene 1.1 ST Array 10707779 - -
          Affymetrix Rat Gene 1.1 ST Array 10707756 - -
          Rat Genome 230 2.0 Array 1373277_at - -
          Rat Genome 230 2.0 Array 1376630_PM_at - -
          Rat Genome 230 2.0 Array 1376630_at - -
          Rat Genome 230 2.0 Array 1373277_PM_at - -
          Affymetrix HT Rat Focus Array 1373277_PM_at - -
          Affymetrix HT Rat Focus Array 1373277_at - -
          Rat Expression Set 230 1373277_at - -
          Rat Expression Set 230 1376630_at - -
          Rat Expression Set 230 1373277_PM_at - -
          Rat Expression Set 230 1376630_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10707779 - -
          Rat Genome U34 Set rc_AI145569_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21112556 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1369090 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-4lm45:80/100.66.75.98:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0