Search
Search

LOC100360933| Gene Symbol: LOC100360933 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001193275.1 | NP_001180204.1 | 4 - 5 | 346 | 88 | |
| XM_008759490.1 | XP_008757712.1 | 1 - 2 | 714 | 88 | ||
| XM_008759491.1 | XP_008757713.1 | 1 - 2 | 202 | 88 | ||
| GenBank mRNA | BF386180.1 | - | 2 - 3 | 88 | ||
Set Membership: |
|