Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01477756_gH
          See other ZFP710 GE Assays ›
          Gene Symbol:
          Zfp710
          Gene Name:
          zinc finger protein 710
          Gene Aliases:
          RGD1306491, Znf710
          Chromosome Location:
          Chr.1: 141763629 - 141870960 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01477756_gH
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Zfp710
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001134563.1 NP_001128035.1 2 - 3 1878 64
          XM_008759507.1 XP_008757729.1 3 - 4 3466 64
          XM_008759508.1 XP_008757730.1 3 - 4 3271 64

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100001849.rn.1

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Probe spans exons

          Panther Classification:

          Molecular Function -

          C2H2 zinc finger transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of transcription from RNA polymerase II promoter
          nucleic acid binding
          transcription cofactor activity
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P557634 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 926815 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 222306 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 290232 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 647711 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P557634 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P557634 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P557634 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P557634 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P1007831 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10708149 - -
          Affymetrix Rat Gene 1.0 ST Array 10708150 - -
          Affymetrix Rat Gene 1.0 ST Array 10708148 - -
          Affymetrix Rat Gene 1.1 ST Array 10708146 - -
          Rat Genome 230 2.0 Array 1389650_at - -
          Rat Genome 230 2.0 Array 1392913_PM_at - -
          Rat Genome 230 2.0 Array 1378928_PM_at - -
          Rat Genome 230 2.0 Array 1392913_at - -
          Rat Genome 230 2.0 Array 1389650_PM_at - -
          Rat Genome 230 2.0 Array 1378928_at - -
          Affymetrix HT Rat Focus Array 1392913_PM_at - -
          Affymetrix HT Rat Focus Array 1378928_at - -
          Affymetrix HT Rat Focus Array 1378928_PM_at - -
          Affymetrix HT Rat Focus Array 1392913_at - -
          Affymetrix Rat Clariom S Assay PSR0100014538.rn.1 TC0100001849.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100014535.rn.1 TC0100001849.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100014536.rn.1 TC0100001849.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100014537.rn.1 TC0100001849.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100014534.rn.1 TC0100001849.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100014539.rn.1 TC0100001849.rn.1 RTA TC
          Rat Expression Set 230 1378928_at - -
          Rat Expression Set 230 1392913_PM_at - -
          Rat Expression Set 230 1378928_PM_at - -
          Rat Expression Set 230 1392913_at - -
          Affymetrix Rat Gene 1.0 ST Array 10708146 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20755865 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1365654 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-h9fps:80/100.66.77.141:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline