Search
Search

Adamts17| Gene Symbol: Adamts17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006229360.2 | XP_006229422.1 | 8 - 9 | 1393 | 59 | |
| XM_218753.8 | XP_218753.6 | 8 - 9 | 1392 | 59 | ||
Set Membership: |
|