Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mphosph10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106340.1 | NP_001099810.1 | 4 - 5 | 1125 | 135 | |
GenBank mRNA | BC161909.1 | - | 4 - 5 | 1098 | 8367841 | 135 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |