Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01478481_m1
          See other ADAM5 GE Assays ›
          Gene Symbol:
          Adam5
          Gene Name:
          a disintegrin and metallopeptidase domain 5
          Gene Aliases:
          tMDCII
          Chromosome Location:
          Chr.16: 72010051 - 72085655 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01478481_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Adam5
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_020303.1 NP_064699.1 9 - 10 904 60
          XM_006253321.2 XP_006253383.1 9 - 10 954 60
          XM_006253322.2 XP_006253384.1 9 - 10 891 60
          GenBank mRNA BC079016.1 - 9 - 10 874 60
          BC091169.1 - 9 - 10 904 7112347 60
          Y11489.1 - 2 - 3 219 60

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1600000836.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Protease 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Adam5

          Panther Classification:

          Molecular Function -

          metalloprotease

          Gene Ontology Categories:

          Function(s) Process(es)

          proteolysis
          metalloendopeptidase activity
          zinc ion binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P159310 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 909542 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 890554 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 793355 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 21649 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 424983 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 663043 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 381253 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 550287 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 477135 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 441185 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 302644 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 6009 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 545353 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 138215 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 804224 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 813853 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 54100 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 790262 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 394547 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 846815 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 300839 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 68817 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 654010 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 85892 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 603443 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 642603 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 826465 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 481689 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 867953 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 150885 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 407492 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 254911 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 168344 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 625876 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 819196 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 760892 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 329954 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 485800 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 307832 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 350709 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 896011 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 101612 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 58764 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 102998 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 140961 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 141518 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 15169 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 115642 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 147924 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 141986 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P159310 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P159310 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P159310 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P16391 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 21925 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 96767 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 13093 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 69700 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 75264 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10788951 - -
          Affymetrix Rat Gene 1.0 ST Array 10788954 - -
          Affymetrix Rat Gene 1.0 ST Array 10788950 - -
          Affymetrix Rat Gene 1.0 ST Array 10788958 - -
          Affymetrix Rat Gene 1.0 ST Array 10788962 - -
          Affymetrix Rat Gene 1.0 ST Array 10788952 - -
          Affymetrix Rat Gene 1.0 ST Array 10788955 - -
          Affymetrix Rat Gene 1.0 ST Array 10788963 - -
          Affymetrix Rat Gene 1.0 ST Array 10788948 - -
          Affymetrix Rat Gene 1.0 ST Array 10788956 - -
          Affymetrix Rat Gene 1.0 ST Array 10788960 - -
          Affymetrix Rat Gene 1.0 ST Array 10788957 - -
          Affymetrix Rat Gene 1.0 ST Array 10788949 - -
          Affymetrix Rat Gene 1.0 ST Array 10788966 - -
          Affymetrix Rat Gene 1.0 ST Array 10788961 - -
          Affymetrix Rat Gene 1.0 ST Array 10788965 - -
          Affymetrix Rat Gene 1.0 ST Array 10788964 - -
          Affymetrix Rat Gene 1.0 ST Array 10788968 - -
          Affymetrix Rat Gene 1.0 ST Array 10788967 - -
          Affymetrix Rat Gene 1.0 ST Array 10788959 - -
          Affymetrix Rat Gene 1.0 ST Array 10788953 - -
          Affymetrix Rat Gene 1.1 ST Array 10788946 - -
          Affymetrix Rat Clariom S Assay PSR1600006388.rn.1 TC1600000836.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1600006400.rn.1 TC1600000836.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1600006392.rn.1 TC1600000836.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1600006395.rn.1 TC1600000836.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1600006390.rn.1 TC1600000836.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1600006385.rn.1 TC1600000836.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1600006401.rn.1 TC1600000836.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1600006393.rn.1 TC1600000836.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1600006389.rn.1 TC1600000836.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1600006398.rn.1 TC1600000836.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1600006391.rn.1 TC1600000836.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1600006396.rn.1 TC1600000836.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1600006397.rn.1 TC1600000836.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1600006399.rn.1 TC1600000836.rn.1 RTA TC
          Affymetrix Rat Gene 1.0 ST Array 10788946 - -
          Rat Genome U34 Set Y11489cds_at - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1350659 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-green-7b6576f7bc-t4kr2:80/100.66.77.141:80.
          git-commit: 6d309ebe7a31b2ceadeb87f43b473eebab110cf3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-2026.03.11-1.0