Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01478566_m1
          See other CDC16 GE Assays ›
          Gene Symbol:
          Cdc16
          Gene Name:
          cell division cycle 16
          Gene Aliases:
          -
          Chromosome Location:
          Chr.16: 81493355 - 81516493 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01478566_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cdc16
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001024744.1 NP_001019915.1 17 - 18 1722 182
          GenBank mRNA BC097498.1 - 17 - 18 1722 7462333 182

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1600000971.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Cdc16

          Panther Classification:

          Molecular Function -

          ubiquitin-protein ligase

          Gene Ontology Categories:

          Function(s)

          cell division
          protein K11-linked ubiquitination

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P999630 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 598810 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 664163 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 164991 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 653421 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 764011 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 484342 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 362087 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 331204 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 454929 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 861190 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 623153 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 145021 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 138909 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 597997 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 550608 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 736846 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 62614 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 149628 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 135867 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 102844 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P999630 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P999630 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P999630 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P999630 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P154367 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 17132 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 62521 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 637 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 55524 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10792717 - -
          Affymetrix Rat Gene 1.0 ST Array 10792725 - -
          Affymetrix Rat Gene 1.0 ST Array 10792714 - -
          Affymetrix Rat Gene 1.0 ST Array 10792718 - -
          Affymetrix Rat Gene 1.0 ST Array 10792715 - -
          Affymetrix Rat Gene 1.0 ST Array 10792721 - -
          Affymetrix Rat Gene 1.0 ST Array 10792727 - -
          Affymetrix Rat Gene 1.0 ST Array 10792720 - -
          Affymetrix Rat Gene 1.0 ST Array 10792726 - -
          Affymetrix Rat Gene 1.0 ST Array 10792731 - -
          Affymetrix Rat Gene 1.0 ST Array 10792724 - -
          Affymetrix Rat Gene 1.0 ST Array 10792729 - -
          Affymetrix Rat Gene 1.0 ST Array 10792716 - -
          Affymetrix Rat Gene 1.0 ST Array 10792728 - -
          Affymetrix Rat Gene 1.0 ST Array 10792730 - -
          Affymetrix Rat Gene 1.0 ST Array 10792723 - -
          Affymetrix Rat Gene 1.0 ST Array 10792722 - -
          Affymetrix Rat Gene 1.0 ST Array 10792719 - -
          Affymetrix Rat Gene 1.1 ST Array 10792713 - -
          Rat Genome 230 2.0 Array 1396172_PM_at - -
          Rat Genome 230 2.0 Array 1399074_at - -
          Rat Genome 230 2.0 Array 1399074_PM_at - -
          Rat Genome 230 2.0 Array 1396172_at - -
          Affymetrix HT Rat Focus Array 1399074_PM_at - -
          Affymetrix HT Rat Focus Array 1396172_at - -
          Affymetrix HT Rat Focus Array 1396172_PM_at - -
          Affymetrix HT Rat Focus Array 1399074_at - -
          Affymetrix Rat Clariom S Assay PSR1600007325.rn.1 TC1600000971.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1600007328.rn.1 TC1600000971.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1600007327.rn.1 TC1600000971.rn.1 RTA TC
          Rat Expression Set 230 1399074_PM_at - -
          Rat Expression Set 230 1399074_at - -
          Rat Expression Set 230 1396172_PM_at - -
          Rat Expression Set 230 1396172_at - -
          Affymetrix Rat Gene 1.0 ST Array 10792713 - -
          Rat Genome U34 Set rc_AI012077_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22351200 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_2039356 - -
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1365596 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline