Search
Search

RGD1564788| Gene Symbol: RGD1564788 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006253254.2 | XP_006253316.1 | 25 - 26 | 3928 | 67 | |
| XM_006253256.2 | XP_006253318.1 | 25 - 26 | 3357 | 67 | ||
| XM_008771311.1 | XP_008769533.1 | 24 - 25 | 4002 | 67 | ||
Set Membership: |
|