Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture and Transfection
    • Chemicals
    • Chromatography
    • Electron Microscopes
    • Lab Plasticware and Supplies
    • Lab Centrifuges
    • Lab Solutions
    • Mass Spectrometers
    • Next Generation Sequencers
    • See all product categories
  • Applications
    • Brands
    • Industrial and Applied Sciences
    • Food and Beverage
    • Forensics
    • Lab Solutions
    • Life Sciences
    • Pharma and Biopharma
    • Biotechnology
    • Clinical and Diagnostics
    • Digital Solutions
    • See all applications
  • Services
    • Lab Instrument and Equipment Services
    • Custom Services
    • Training Services
    • Financial and Leasing Services
    • Enterprise Level Lab Informatics
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Cell Biology Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • Contact Us
    • Product Documentation
    • Knowledge Base and Product FAQs
    • Learning Centers
    • Supply Center
    • eProcurement Solutions
    • Lab Instrument and Equipment Support
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01478742_m1
          See other POTEG GE Assays ›
          Gene Symbol:
          Poteg
          Gene Name:
          POTE ankyrin domain family, member G
          Gene Aliases:
          -
          Chromosome Location:
          Chr.16: 68738819 - 68832911 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01478742_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Poteg
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001014238.1 NP_001014260.1 4 - 5 761 148
          XM_008771330.1 XP_008769552.1 4 - 5 810 148
          GenBank mRNA BC079270.1 - 4 - 5 761 7133714 148

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1600001829.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Poteg

          Panther Classification:

          Molecular Function -

          membrane traffic protein

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P139644 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 437208 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 926323 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 389059 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 509765 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 632843 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 100365 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 489050 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 529620 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 109005 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 431950 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 463394 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 667095 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 755662 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 47884 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 856041 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 775676 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 113748 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P139644 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P139644 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P139644 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 18553 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 31179 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 55274 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10792146 - -
          Affymetrix Rat Gene 1.0 ST Array 10792148 - -
          Affymetrix Rat Gene 1.0 ST Array 10792142 - -
          Affymetrix Rat Gene 1.0 ST Array 10792149 - -
          Affymetrix Rat Gene 1.0 ST Array 10792145 - -
          Affymetrix Rat Gene 1.0 ST Array 10792147 - -
          Affymetrix Rat Gene 1.0 ST Array 10792150 - -
          Affymetrix Rat Gene 1.0 ST Array 10792144 - -
          Affymetrix Rat Gene 1.0 ST Array 10792143 - -
          Affymetrix Rat Gene 1.0 ST Array 10792140 - -
          Affymetrix Rat Gene 1.1 ST Array 10792139 - -
          Affymetrix Rat Clariom S Assay PSR1600013483.rn.1 TC1600001829.rn.1 RTA TC
          Affymetrix Rat Gene 1.0 ST Array 10792139 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22044067 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Quick Order
          • eProcurement
          • Supply Center
          • Order Status
          • Chemicals
          • India Mobile App
          • Government eMarketplace
          Support Plus Icon Minus Icon
          • Order Support
          • Training
          • Contact Us
          • Report a Site Issue
          • Instrument Management
          Resources Plus Icon Minus Icon
          • Product Selection Guides
          • Mobile & Desktop Apps
          • Webinars
          • Blog 
          • Social Media
          • New Products
          • Promotions
          • Shared Lists
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          India flag icon
          India

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline