Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01478946_m1
          See other LPXN GE Assays ›
          Gene Symbol:
          Lpxn
          Gene Name:
          leupaxin
          Gene Aliases:
          -
          Chromosome Location:
          Chr.1: 229642437 - 229674129 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01478946_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Lpxn
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001009649.1 NP_001009649.1 7 - 8 810 74
          GenBank mRNA BC088217.1 - 7 - 8 810 7384057 74
          FQ230386.1 - 7 - 8 74

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100003266.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Lpxn

          Panther Classification:

          Molecular Function -

          actin or actin-binding cytoskeletal protein

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of cell adhesion
          biological_process
          regulation of cell adhesion mediated by integrin
          negative regulation of B cell receptor signaling pathway
          regulation of nucleic acid-templated transcription
          molecular_function
          transcription cofactor activity
          zinc ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P23014 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 376396 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 825400 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 558425 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 181889 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 171267 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 158257 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 930007 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 732876 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 620693 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 245737 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 538155 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 269514 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 306107 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 813716 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 570050 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 27302 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 792946 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 176748 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 262330 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 225462 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 520789 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 281428 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 264999 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 259334 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 292679 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 373676 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 377069 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 171964 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 15826 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 53198 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 50437 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P23014 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P23014 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P23014 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P23014 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 77216 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 38988 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 72521 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10714142 - -
          Affymetrix Rat Gene 1.0 ST Array 10714141 - -
          Affymetrix Rat Gene 1.0 ST Array 10714143 - -
          Affymetrix Rat Gene 1.0 ST Array 10714140 - -
          Affymetrix Rat Gene 1.0 ST Array 10714137 - -
          Affymetrix Rat Gene 1.0 ST Array 10714138 - -
          Affymetrix Rat Gene 1.0 ST Array 10714146 - -
          Affymetrix Rat Gene 1.0 ST Array 10714144 - -
          Affymetrix Rat Gene 1.0 ST Array 10714139 - -
          Affymetrix Rat Gene 1.0 ST Array 10714145 - -
          Affymetrix Rat Gene 1.1 ST Array 10714136 - -
          Rat Genome 230 2.0 Array 1378633_PM_at - -
          Rat Genome 230 2.0 Array 1378633_at - -
          Affymetrix HT Rat Focus Array 1378633_PM_at - -
          Affymetrix HT Rat Focus Array 1378633_at - -
          Affymetrix Rat Clariom S Assay PSR0100026970.rn.1 TC0100003266.rn.1 RTA TC
          Rat Expression Set 230 1378633_at - -
          Rat Expression Set 230 1378633_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10714136 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21526742 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1372207 - -
          RatRef-12 Expression BeadChip ILMN_54230 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-4lm45:80/100.66.75.98:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0