Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01479091_g1
          See other SYVN1 GE Assays ›
          Gene Symbol:
          Syvn1
          Gene Name:
          synovial apoptosis inhibitor 1, synoviolin
          Gene Aliases:
          RGD1310488
          Chromosome Location:
          Chr.1: 221409265 - 221416225 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01479091_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Syvn1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001100739.1 NP_001094209.1 14 - 15 1681 124
          XM_006230852.2 XP_006230914.1 14 - 15 2123 124
          GenBank mRNA BC099172.1 - 3 - 4 472 124
          BC166727.1 - 14 - 15 1681 7933989 124

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100003084.rn.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          in utero embryonic development
          protein N-linked glycosylation via asparagine
          ER-associated ubiquitin-dependent protein catabolic process
          negative regulation of endoplasmic reticulum stress-induced intrinsic apoptotic signaling pathway
          zinc ion binding
          acid-amino acid ligase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P871622 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 391251 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 760436 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 388386 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 45637 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 88556 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 76649 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P871622 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P871622 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P871622 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P126134 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P871622 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10713171 - -
          Affymetrix Rat Gene 1.0 ST Array 10713161 - -
          Affymetrix Rat Gene 1.0 ST Array 10713165 - -
          Affymetrix Rat Gene 1.0 ST Array 10713174 - -
          Affymetrix Rat Gene 1.0 ST Array 10713170 - -
          Affymetrix Rat Gene 1.0 ST Array 10713162 - -
          Affymetrix Rat Gene 1.0 ST Array 10713175 - -
          Affymetrix Rat Gene 1.0 ST Array 10713169 - -
          Affymetrix Rat Gene 1.0 ST Array 10713164 - -
          Affymetrix Rat Gene 1.0 ST Array 10713173 - -
          Affymetrix Rat Gene 1.0 ST Array 10713172 - -
          Affymetrix Rat Gene 1.0 ST Array 10713163 - -
          Affymetrix Rat Gene 1.0 ST Array 10713167 - -
          Affymetrix Rat Gene 1.0 ST Array 10713166 - -
          Affymetrix Rat Gene 1.0 ST Array 10713168 - -
          Affymetrix Rat Gene 1.1 ST Array 10713160 - -
          Rat Genome 230 2.0 Array 1371827_at - -
          Rat Genome 230 2.0 Array 1371827_PM_at - -
          Affymetrix HT Rat Focus Array 1371827_PM_at - -
          Affymetrix HT Rat Focus Array 1371827_at - -
          Affymetrix Rat Clariom S Assay PSR0100025039.rn.1 TC0100003084.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100025041.rn.1 TC0100003084.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100025040.rn.1 TC0100003084.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100025036.rn.1 TC0100003084.rn.1 RTA TC
          Rat Expression Set 230 1371827_PM_at - -
          Rat Expression Set 230 1371827_at - -
          Affymetrix Rat Gene 1.0 ST Array 10713160 - -
          Rat Genome U34 Set rc_AA946430_at - -
          Rat Genome U34 Set rc_AA858449_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21679497 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1351199 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-j77tp:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline