Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01479150_g1
          See other SMARCA2 GE Assays ›
          Gene Symbol:
          Smarca2
          Gene Name:
          SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily a, member 2
          Gene Aliases:
          -
          Chromosome Location:
          Chr.1: 244615814 - 244783736 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01479150_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Smarca2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001004446.1 NP_001004446.1 33 - 34 4912 58
          XM_006231227.2 XP_006231289.1 32 - 33 4876 58
          XM_006231229.2 XP_006231291.1 7 - 8 977 58
          XM_008760331.1 XP_008758553.1 33 - 34 5190 58
          XM_008760332.1 XP_008758554.1 34 - 35 5238 58
          XM_008760333.1 XP_008758555.1 34 - 35 4978 58
          XM_008760334.1 XP_008758556.1 33 - 34 5010 58
          XM_008760335.1 XP_008758557.1 33 - 34 4946 58
          XM_008760336.1 XP_008758558.1 33 - 34 5258 58
          XM_008760337.1 XP_008758559.1 32 - 33 4808 58
          XM_008760338.1 XP_008758560.1 31 - 32 4444 58
          XM_008760339.1 XP_008758561.1 7 - 8 879 58
          GenBank mRNA AY643746.1 - 33 - 34 4912 58

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Smarca2

          Panther Classification:

          Molecular Function -

          DNA helicase

          Gene Ontology Categories:

          Function(s) Process(es)

          chromatin remodeling
          spermatid development
          biological_process
          metabolic process
          negative regulation of cell proliferation
          negative regulation of cell growth
          aortic smooth muscle cell differentiation
          negative regulation of transcription, DNA-templated
          positive regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          RNA polymerase II transcription coactivator activity
          chromatin binding
          ATP binding
          ATPase activity
          histone binding
          transcription regulatory region DNA binding

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P735279 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 538995 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 614859 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 272381 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 364840 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 224786 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 344217 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 706225 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 751361 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 189543 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 968098 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 940956 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 522526 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 138870 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 295707 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 940714 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 746329 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 966592 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 624666 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 243940 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 201781 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 484229 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 922865 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 874731 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 269924 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 962443 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 245767 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 630490 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 257816 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 838635 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 209509 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 234540 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 234064 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 268373 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 703125 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 770418 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 418989 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 39195 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 803121 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 560213 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 111761 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 834108 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 241996 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 418346 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 362637 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 397546 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 956451 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 389840 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 932422 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 6243 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 50245 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 444429 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 382748 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 950897 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 731514 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 390454 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 111561 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 954055 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 735862 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 837131 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 583962 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 486807 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 294540 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 423321 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 543315 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 560477 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 179717 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 570880 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 394108 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 949563 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 277519 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 94113 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 923341 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 960741 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 750438 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 337093 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 543365 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 237719 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 553621 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 17269 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 327198 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 337939 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 621364 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 125538 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 748961 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 755605 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 178057 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 409389 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 352728 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 858378 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 743262 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 969241 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 18469 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 254889 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 370555 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 456656 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 416503 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 17091 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 320498 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 955718 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 878250 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 250433 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 613775 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 602130 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 725451 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 507236 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 87346 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 228040 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 9346 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 547190 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 342566 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 361559 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 831652 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 605710 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 261422 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 306463 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 31753 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 453260 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 37377 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 247824 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 394932 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 705795 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 582720 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 681693 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 206795 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 516154 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 545697 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 634183 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 714407 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 334160 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 715085 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 916939 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 176426 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 717101 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 223621 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 42430 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 29095 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 60496 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 31717 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 55365 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 12695 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 28536 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 177069 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 91845 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 142380 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 102141 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 129037 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 3209 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 54271 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 68982 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 169224 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 148245 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 104897 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 17843 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 111723 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P735279 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P735279 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P735279 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P1021682 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 77102 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 26395 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 97825 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 104942 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 42996 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 54528 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 54165 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 80028 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 11097 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 23144 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 55763 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10714541 - -
          Affymetrix Rat Gene 1.0 ST Array 10714544 - -
          Affymetrix Rat Gene 1.0 ST Array 10714570 - -
          Affymetrix Rat Gene 1.0 ST Array 10714545 - -
          Affymetrix Rat Gene 1.0 ST Array 10714561 - -
          Affymetrix Rat Gene 1.0 ST Array 10714571 - -
          Affymetrix Rat Gene 1.0 ST Array 10714548 - -
          Affymetrix Rat Gene 1.0 ST Array 10714555 - -
          Affymetrix Rat Gene 1.0 ST Array 10714567 - -
          Affymetrix Rat Gene 1.0 ST Array 10714562 - -
          Affymetrix Rat Gene 1.0 ST Array 10714556 - -
          Affymetrix Rat Gene 1.0 ST Array 10714552 - -
          Affymetrix Rat Gene 1.0 ST Array 10714542 - -
          Affymetrix Rat Gene 1.0 ST Array 10714568 - -
          Affymetrix Rat Gene 1.0 ST Array 10714559 - -
          Affymetrix Rat Gene 1.0 ST Array 10714547 - -
          Affymetrix Rat Gene 1.0 ST Array 10714546 - -
          Affymetrix Rat Gene 1.0 ST Array 10714564 - -
          Affymetrix Rat Gene 1.0 ST Array 10714560 - -
          Affymetrix Rat Gene 1.0 ST Array 10714569 - -
          Affymetrix Rat Gene 1.0 ST Array 10714557 - -
          Affymetrix Rat Gene 1.0 ST Array 10714565 - -
          Affymetrix Rat Gene 1.0 ST Array 10714558 - -
          Affymetrix Rat Gene 1.0 ST Array 10714549 - -
          Affymetrix Rat Gene 1.0 ST Array 10714538 - -
          Affymetrix Rat Gene 1.0 ST Array 10714540 - -
          Affymetrix Rat Gene 1.0 ST Array 10714553 - -
          Affymetrix Rat Gene 1.0 ST Array 10869556 - -
          Affymetrix Rat Gene 1.0 ST Array 10714566 - -
          Affymetrix Rat Gene 1.0 ST Array 10714550 - -
          Affymetrix Rat Gene 1.1 ST Array 10869555 - -
          Affymetrix Rat Gene 1.1 ST Array 10714537 - -
          Rat Genome 230 2.0 Array 1376337_PM_at - -
          Rat Genome 230 2.0 Array 1385074_PM_at - -
          Rat Genome 230 2.0 Array 1377312_PM_at - -
          Rat Genome 230 2.0 Array 1377312_at - -
          Rat Genome 230 2.0 Array 1394066_at - -
          Rat Genome 230 2.0 Array 1397165_PM_at - -
          Rat Genome 230 2.0 Array 1397165_at - -
          Rat Genome 230 2.0 Array 1394066_PM_at - -
          Rat Genome 230 2.0 Array 1385074_at - -
          Rat Genome 230 2.0 Array 1376337_at - -
          Affymetrix HT Rat Focus Array 1385074_at - -
          Affymetrix HT Rat Focus Array 1376337_at - -
          Affymetrix HT Rat Focus Array 1394066_at - -
          Affymetrix HT Rat Focus Array 1394066_PM_at - -
          Affymetrix HT Rat Focus Array 1376337_PM_at - -
          Affymetrix HT Rat Focus Array 1385074_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0100027764.rn.1 TC0100003413.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100027767.rn.1 TC0100003413.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100027768.rn.1 TC0100003413.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100027765.rn.1 TC0100003413.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100027766.rn.1 TC0100003413.rn.1 RTA TC
          Rat Expression Set 230 1394066_at - -
          Rat Expression Set 230 1376337_PM_at - -
          Rat Expression Set 230 1376337_at - -
          Rat Expression Set 230 1394066_PM_at - -
          Rat Expression Set 230 1385074_PM_at - -
          Rat Expression Set 230 1385074_at - -
          Affymetrix Rat Gene 1.0 ST Array 10714537 - -
          Rat Genome U34 Set rc_AA875609_at - -
          Rat Genome U34 Set rc_AA899635_at - -
          Rat Genome U34 Set rc_AA963961_at - -
          Rat Genome U34 Set rc_AI170385_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21183903 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline