Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01479181_m1
          See other SCYL1 GE Assays ›
          Gene Symbol:
          Scyl1
          Gene Name:
          SCY1-like 1 (S. cerevisiae)
          Gene Aliases:
          -
          Chromosome Location:
          Chr.1: 221115992 - 221129666 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01479181_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Scyl1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001011938.1 NP_001011938.1 8 - 9 1255 123
          XM_006230749.2 XP_006230811.2 8 - 9 1317 123
          GenBank mRNA BC087141.1 - 8 - 9 1255 7127562 123
          FQ217187.1 - 8 - 9 1305 123

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100007135.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Protein Kinase Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Scyl1

          Panther Classification:

          Molecular Function -

          non-receptor serine/threonine protein kinase

          Gene Ontology Categories:

          Function(s) Process(es)

          protein phosphorylation
          retrograde vesicle-mediated transport, Golgi to ER
          protein kinase activity
          ATP binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P142056 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 919130 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 289776 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 167368 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 432963 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 375793 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 48727 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 906656 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 939978 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 525386 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 115019 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 19648 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P142056 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P142056 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P142056 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P148807 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 90949 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10728019 - -
          Affymetrix Rat Gene 1.0 ST Array 10728015 - -
          Affymetrix Rat Gene 1.0 ST Array 10728017 - -
          Affymetrix Rat Gene 1.0 ST Array 10728024 - -
          Affymetrix Rat Gene 1.0 ST Array 10728023 - -
          Affymetrix Rat Gene 1.0 ST Array 10728016 - -
          Affymetrix Rat Gene 1.0 ST Array 10728013 - -
          Affymetrix Rat Gene 1.0 ST Array 10728018 - -
          Affymetrix Rat Gene 1.0 ST Array 10728011 - -
          Affymetrix Rat Gene 1.0 ST Array 10728021 - -
          Affymetrix Rat Gene 1.0 ST Array 10728022 - -
          Affymetrix Rat Gene 1.0 ST Array 10728027 - -
          Affymetrix Rat Gene 1.0 ST Array 10728014 - -
          Affymetrix Rat Gene 1.0 ST Array 10728020 - -
          Affymetrix Rat Gene 1.0 ST Array 10728025 - -
          Affymetrix Rat Gene 1.0 ST Array 10728026 - -
          Affymetrix Rat Gene 1.0 ST Array 10728012 - -
          Affymetrix Rat Gene 1.1 ST Array 10728010 - -
          Rat Genome 230 2.0 Array 1399024_at - -
          Rat Genome 230 2.0 Array 1399024_PM_at - -
          Rat Genome 230 2.0 Array 1382635_PM_at - -
          Rat Genome 230 2.0 Array 1382635_at - -
          Affymetrix HT Rat Focus Array 1399024_at - -
          Affymetrix HT Rat Focus Array 1382635_at - -
          Affymetrix HT Rat Focus Array 1399024_PM_at - -
          Affymetrix HT Rat Focus Array 1382635_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0100057159.rn.1 TC0100007135.rn.1 RTA TC
          Rat Expression Set 230 1399024_PM_at - -
          Rat Expression Set 230 1399024_at - -
          Rat Expression Set 230 1382635_at - -
          Rat Expression Set 230 1382635_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10728010 - -
          Rat Genome U34 Set rc_AA957167_f_at - -
          Rat Genome U34 Set rc_AI180441_at - -
          Rat Genome U34 Set rc_AI234887_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21001933 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1376547 - -
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1375876 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline