Search
Search
| Gene Symbol: Pkp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106315.1 | NP_001099785.1 | 6 - 7 | 1459 | 88 | |
| XM_006230531.2 | XP_006230593.1 | 5 - 6 | 2627 | 88 | ||
| XM_008759982.1 | XP_008758204.1 | 5 - 6 | 2627 | 88 | ||
Set Membership: |
|