Search
Search
| Gene Symbol: Ppp6r3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001287134.1 | NP_001274063.1 | 8 - 9 | 1077 | 70 | |
| XM_008760121.1 | XP_008758343.1 | 8 - 9 | 1631 | 70 | ||
| XM_008760122.1 | XP_008758344.1 | 8 - 9 | 1631 | 70 | ||
| XM_008760123.1 | XP_008758345.1 | 8 - 9 | 1631 | 70 | ||
| XM_008760124.1 | XP_008758346.1 | 8 - 9 | 1631 | 70 | ||
| XM_008760125.1 | XP_008758347.1 | 8 - 9 | 1631 | 70 | ||
| XM_008760126.1 | XP_008758348.1 | 8 - 9 | 1631 | 70 | ||
| XM_008760127.1 | XP_008758349.1 | 8 - 9 | 1631 | 70 | ||
| XM_008760128.1 | XP_008758350.1 | 9 - 10 | 1246 | 70 | ||
| XM_008760129.1 | XP_008758351.1 | 8 - 9 | 1098 | 70 | ||
| XM_008760130.1 | XP_008758352.1 | 8 - 9 | 2267 | 70 | ||
| XM_008760131.1 | XP_008758353.1 | 8 - 9 | 1098 | 70 | ||
Set Membership: |
|