Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01479319_g1
          See other RFK GE Assays ›
          Gene Symbol:
          Rfk
          Gene Name:
          riboflavin kinase
          Gene Aliases:
          -
          Chromosome Location:
          Chr.1: 236494221 - 236501733 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01479319_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rfk
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001014106.1 NP_001014128.1 2 - 3 491 132
          GenBank mRNA BC079125.1 - 2 - 3 491 7127049 132
          FQ219206.1 - 2 - 3 459 132
          FQ223869.1 - 2 - 3 504 132

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Rfk

          Gene Ontology Categories:

          Function(s) Process(es)

          apoptotic process
          riboflavin biosynthetic process
          phosphorylation
          positive regulation of NAD(P)H oxidase activity
          reactive oxygen species metabolic process
          riboflavin kinase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P110971 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P143307 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P110966 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 940831 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 211774 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 555448 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 350034 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 156667 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 103699 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P110971 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P110966 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P143307 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P110966 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P110971 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P143307 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P110971 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P110966 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P143307 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P326001 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10714250 - -
          Affymetrix Rat Gene 1.0 ST Array 10714252 - -
          Affymetrix Rat Gene 1.0 ST Array 10934663 - -
          Affymetrix Rat Gene 1.0 ST Array 10847719 - -
          Affymetrix Rat Gene 1.0 ST Array 10714251 - -
          Affymetrix Rat Gene 1.0 ST Array 10714253 - -
          Affymetrix Rat Gene 1.1 ST Array 10847718 - -
          Affymetrix Rat Gene 1.1 ST Array 10934662 - -
          Affymetrix Rat Gene 1.1 ST Array 10714249 - -
          Rat Genome 230 2.0 Array 1382331_PM_at - -
          Rat Genome 230 2.0 Array 1372671_at - -
          Rat Genome 230 2.0 Array 1372671_PM_at - -
          Rat Genome 230 2.0 Array 1382331_at - -
          Affymetrix HT Rat Focus Array 1382331_PM_at - -
          Affymetrix HT Rat Focus Array 1372671_PM_at - -
          Affymetrix HT Rat Focus Array 1372671_at - -
          Affymetrix HT Rat Focus Array 1382331_at - -
          Affymetrix Rat Clariom S Assay PSR0100059624.rn.1 TC0100007375.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100059625.rn.1 TC0100007375.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100059622.rn.1 TC0100007375.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100059627.rn.1 TC0100007375.rn.1 RTA TC
          Rat Expression Set 230 1372671_at - -
          Rat Expression Set 230 1372671_PM_at - -
          Rat Expression Set 230 1382331_PM_at - -
          Rat Expression Set 230 1382331_at - -
          Affymetrix Rat Gene 1.0 ST Array 10714249 - -
          Affymetrix Rat Gene 1.0 ST Array 10847718 - -
          Rat Genome U34 Set rc_AA859848_at - -
          Rat Genome U34 Set rc_AA849030_at - -
          Rat Genome U34 Set rc_AI237193_at - -
          Rat Genome U34 Set rc_AI180421_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22362420 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1368534 - -
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1649985 - -
          RatRef-12 Expression BeadChip ILMN_67885 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline