Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01479669_m1
          See other TOLLIP GE Assays ›
          Gene Symbol:
          Tollip
          Gene Name:
          toll interacting protein
          Gene Aliases:
          -
          Chromosome Location:
          Chr.1: 214823993 - 214844861 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01479669_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tollip
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001109668.1 NP_001103138.1 5 - 6 799 64
          XM_006230607.2 XP_006230669.1 6 - 7 880 64
          XM_008760003.1 XP_008758225.1 5 - 6 744 64
          GenBank mRNA BC133067.1 - 5 - 6 685 7193007 64

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100006958.rn.1

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          autophagy
          inflammatory response
          signal transduction
          phosphorylation
          epithelial cell differentiation
          positive regulation of protein sumoylation
          protein localization to endosome
          innate immune response
          interleukin-1, Type I receptor binding
          kinase binding
          ubiquitin conjugating enzyme binding
          ubiquitin protein ligase binding
          SUMO binding
          Toll-like receptor binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P262569 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P834980 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 57365 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 245286 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 637434 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 730829 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 789606 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 164811 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 569370 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 33948 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 704501 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 890896 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 258429 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 148706 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 360887 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 34760 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 863329 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 28857 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 174163 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 44192 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P262569 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P834980 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P834980 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P262569 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P262569 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P834980 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P262569 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P834980 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 72089 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10726920 - -
          Affymetrix Rat Gene 1.0 ST Array 10726921 - -
          Affymetrix Rat Gene 1.0 ST Array 10726918 - -
          Affymetrix Rat Gene 1.0 ST Array 10726919 - -
          Affymetrix Rat Gene 1.0 ST Array 10726923 - -
          Affymetrix Rat Gene 1.0 ST Array 10726922 - -
          Affymetrix Rat Gene 1.1 ST Array 10726916 - -
          Rat Genome 230 2.0 Array 1399052_at - -
          Rat Genome 230 2.0 Array 1380808_PM_at - -
          Rat Genome 230 2.0 Array 1378572_at - -
          Rat Genome 230 2.0 Array 1380808_at - -
          Rat Genome 230 2.0 Array 1399052_PM_at - -
          Rat Genome 230 2.0 Array 1378572_PM_at - -
          Affymetrix HT Rat Focus Array 1399052_at - -
          Affymetrix HT Rat Focus Array 1378572_at - -
          Affymetrix HT Rat Focus Array 1378572_PM_at - -
          Affymetrix HT Rat Focus Array 1399052_PM_at - -
          Affymetrix HT Rat Focus Array 1380808_at - -
          Affymetrix HT Rat Focus Array 1380808_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0100054973.rn.1 TC0100006958.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100054971.rn.1 TC0100006958.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100054972.rn.1 TC0100006958.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100054974.rn.1 TC0100006958.rn.1 RTA TC
          Rat Expression Set 230 1399052_PM_at - -
          Rat Expression Set 230 1399052_at - -
          Rat Expression Set 230 1378572_PM_at - -
          Rat Expression Set 230 1380808_PM_at - -
          Rat Expression Set 230 1380808_at - -
          Rat Expression Set 230 1378572_at - -
          Affymetrix Rat Gene 1.0 ST Array 10726916 - -
          Rat Genome U34 Set rc_AA956943_at - -
          Rat Genome U34 Set rc_AI227705_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21352983 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1371790 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline