Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01479681_m1
          See other PYGM GE Assays ›
          Gene Symbol:
          Pygm
          Gene Name:
          phosphorylase, glycogen, muscle
          Gene Aliases:
          Muscpho
          Chromosome Location:
          Chr.1: 221749672 - 221771142 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01479681_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Pygm
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_012638.1 NP_036770.1 5 - 6 701 63
          XM_006230678.2 XP_006230740.1 5 - 6 741 63
          GenBank mRNA BC161897.1 - 5 - 6 696 8366912 63
          L10669.1 - 5 - 6 652 63

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100003108.rn.1

          Set Membership:

          Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Panther Classification:

          Molecular Function -

          glycosyltransferase transferase metabolite interconversion enzyme

          Gene Ontology Categories:

          Function(s) Process(es)

          response to hypoxia
          glycogen metabolic process
          glycogen catabolic process
          cellular calcium ion homeostasis
          response to organic substance
          response to cAMP
          drug binding
          glycogen phosphorylase activity
          AMP binding
          pyridoxal phosphate binding
          carbohydrate binding
          protein homodimerization activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P523853 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 729599 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 245523 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 105581 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 426591 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 55321 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 807592 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 504233 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 130074 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 99275 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 847689 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 620795 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 473486 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 146851 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 171943 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P523853 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P523853 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P523853 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P523853 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 79621 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 19981 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10713396 - -
          Affymetrix Rat Gene 1.0 ST Array 10713387 - -
          Affymetrix Rat Gene 1.0 ST Array 10713400 - -
          Affymetrix Rat Gene 1.0 ST Array 10713392 - -
          Affymetrix Rat Gene 1.0 ST Array 10713390 - -
          Affymetrix Rat Gene 1.0 ST Array 10713398 - -
          Affymetrix Rat Gene 1.0 ST Array 10713383 - -
          Affymetrix Rat Gene 1.0 ST Array 10713399 - -
          Affymetrix Rat Gene 1.0 ST Array 10713391 - -
          Affymetrix Rat Gene 1.0 ST Array 10713388 - -
          Affymetrix Rat Gene 1.0 ST Array 10713397 - -
          Affymetrix Rat Gene 1.0 ST Array 10713389 - -
          Affymetrix Rat Gene 1.0 ST Array 10713393 - -
          Affymetrix Rat Gene 1.0 ST Array 10713401 - -
          Affymetrix Rat Gene 1.0 ST Array 10713384 - -
          Affymetrix Rat Gene 1.0 ST Array 10713395 - -
          Affymetrix Rat Gene 1.0 ST Array 10713385 - -
          Affymetrix Rat Gene 1.0 ST Array 10713394 - -
          Affymetrix Rat Gene 1.0 ST Array 10713386 - -
          Affymetrix Rat Gene 1.1 ST Array 10713382 - -
          Rat Genome 230 2.0 Array 1383072_at - -
          Rat Genome 230 2.0 Array 1370982_PM_at - -
          Rat Genome 230 2.0 Array 1370982_at - -
          Rat Genome 230 2.0 Array 1383072_PM_at - -
          Rat Genome 230 2.0 Array 1375476_at - -
          Rat Genome 230 2.0 Array 1375476_PM_at - -
          Affymetrix HT Rat Focus Array 1383072_PM_at - -
          Affymetrix HT Rat Focus Array 1375476_at - -
          Affymetrix HT Rat Focus Array 1370982_PM_at - -
          Affymetrix HT Rat Focus Array 1375476_PM_at - -
          Affymetrix HT Rat Focus Array 1383072_at - -
          Affymetrix HT Rat Focus Array 1370982_at - -
          Affymetrix Rat Clariom S Assay PSR0100025376.rn.1 TC0100003108.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100025377.rn.1 TC0100003108.rn.1 RTA TC
          Rat Expression Set 230 1375476_PM_at - -
          Rat Expression Set 230 1370982_at - -
          Rat Expression Set 230 1370982_PM_at - -
          Rat Expression Set 230 1383072_PM_at - -
          Rat Expression Set 230 1383072_at - -
          Rat Expression Set 230 1375476_at - -
          Affymetrix Rat Gene 1.0 ST Array 10713382 - -
          Rat Genome U34 Set L10669_at - -
          Rat Genome U34 Set L10669_g_at - -
          Rat Genome U34 Set rc_AI231823_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21255269 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1370766 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline