Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01479927_g1
          See other RPLP2 GE Assays ›
          Gene Symbol:
          Rplp2
          Gene Name:
          ribosomal protein, large P2
          Gene Aliases:
          RP2
          Chromosome Location:
          Chr.1: 214428280 - 214430551 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01479927_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rplp2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001030021.1 NP_001025192.1 3 - 4 217 130
          GenBank mRNA BC099697.1 - 3 - 4 217 7934014 130
          FQ217138.1 - 3 - 4 230 130
          FQ217658.1 - 3 - 4 231 130
          FQ221181.1 - 3 - 4 230 130
          FQ221478.1 - 3 - 4 233 130
          FQ221584.1 - 3 - 4 229 130
          FQ221984.1 - 3 - 4 231 130
          FQ222461.1 - 3 - 4 229 130
          FQ222498.1 - 3 - 4 230 130
          FQ223422.1 - 3 - 4 232 130
          FQ228678.1 - 3 - 4 231 130
          X15098.1 - 3 - 4 231 130

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Endogenous Controls Detect Genomic DNA Recommended Assay Rattus norvegicus Rat Chemokine Panel Rat Endogenous Controls Inventoried Rat_Endogenous_Controls Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Rplp2

          Panther Classification:

          Molecular Function -

          ribosomal protein

          Gene Ontology Categories:

          Function(s) Process(es)

          translational elongation
          structural constituent of ribosome

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P001535 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 731488 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 528838 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P001535 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P001535 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P001535 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P531848 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10776810 - -
          Affymetrix Rat Gene 1.0 ST Array 10781776 - -
          Affymetrix Rat Gene 1.0 ST Array 10712311 - -
          Affymetrix Rat Gene 1.0 ST Array 10712312 - -
          Affymetrix Rat Gene 1.0 ST Array 10712310 - -
          Affymetrix Rat Gene 1.0 ST Array 10712314 - -
          Affymetrix Rat Gene 1.0 ST Array 10712313 - -
          Affymetrix Rat Gene 1.1 ST Array 10781775 - -
          Affymetrix Rat Gene 1.1 ST Array 10776809 - -
          Affymetrix Rat Gene 1.1 ST Array 10712309 - -
          Rat Genome 230 2.0 Array 1371340_PM_at - -
          Rat Genome 230 2.0 Array 1371340_at - -
          Affymetrix Rat Clariom S Assay PSR0100023042.rn.1 TC0100002899.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100023044.rn.1 TC0100002899.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100023045.rn.1 TC0100002899.rn.1 RTA TC
          Rat Expression Set 230 1371340_at - -
          Rat Expression Set 230 1371340_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10781775 - -
          Affymetrix Rat Gene 1.0 ST Array 10776809 - -
          Affymetrix Rat Gene 1.0 ST Array 10712309 - -
          Rat Genome U34 Set X55153mRNA_s_at - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1375860 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-4lm45:80/100.66.75.98:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0