Search
Search
| Gene Symbol: Nkg7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133540.1 | NP_598224.1 | 1 - 2 | 303 | 101 | |
| GenBank mRNA | AF082535.1 | - | 1 - 2 | 303 | 101 | |
| FQ222255.1 | - | 1 - 2 | 309 | 101 | ||
| FQ226323.1 | - | 1 - 2 | 309 | 101 | ||
Set Membership: |
|