Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01480215_m1
          See other SPTBN4 GE Assays ›
          Gene Symbol:
          Sptbn4
          Gene Name:
          spectrin, beta, non-erythrocytic 4
          Gene Aliases:
          Spnb4
          Chromosome Location:
          Chr.1: 84168492 - 84254686 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01480215_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Sptbn4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001100845.2 NP_001094315.2 20 - 21 4478 57
          XM_008759140.1 XP_008757362.1 20 - 21 4519 57
          XM_008759141.1 XP_008757363.1 20 - 21 4519 57

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100005042.rn.1

          Set Membership:

          Amplicon length less than or equal to 70 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of sodium ion transport
          axonogenesis
          sensory perception of sound
          adult walking behavior
          biological_process
          fertilization
          negative regulation of heart rate
          transmission of nerve impulse
          central nervous system projection neuron axonogenesis
          reproductive process
          adult behavior
          regulation of peptidyl-serine phosphorylation
          cellular protein localization
          positive regulation of multicellular organism growth
          clustering of voltage-gated sodium channels
          cardiac conduction
          establishment of protein localization to plasma membrane
          molecular_function
          actin binding
          structural constituent of cytoskeleton
          phospholipid binding
          phosphatase binding
          ankyrin binding
          spectrin binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P125676 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P125676 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P125676 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P125676 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P482710 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P20007 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10720061 - -
          Affymetrix Rat Gene 1.0 ST Array 10720060 - -
          Affymetrix Rat Gene 1.0 ST Array 10720064 - -
          Affymetrix Rat Gene 1.0 ST Array 10720062 - -
          Affymetrix Rat Gene 1.0 ST Array 10720059 - -
          Affymetrix Rat Gene 1.0 ST Array 10720057 - -
          Affymetrix Rat Gene 1.0 ST Array 10720058 - -
          Affymetrix Rat Gene 1.0 ST Array 10720063 - -
          Affymetrix Rat Gene 1.1 ST Array 10720056 - -
          Rat Genome 230 2.0 Array 1376564_at - -
          Rat Genome 230 2.0 Array 1376564_PM_at - -
          Affymetrix HT Rat Focus Array 1376564_PM_at - -
          Affymetrix HT Rat Focus Array 1376564_at - -
          Affymetrix Rat Clariom S Assay PSR0100040158.rn.1 TC0100005042.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100040161.rn.1 TC0100005042.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100040162.rn.1 TC0100005042.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100040165.rn.1 TC0100005042.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100040163.rn.1 TC0100005042.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100040164.rn.1 TC0100005042.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100040159.rn.1 TC0100005042.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100040160.rn.1 TC0100005042.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100040166.rn.1 TC0100005042.rn.1 RTA TC
          Rat Expression Set 230 1376564_at - -
          Rat Expression Set 230 1376564_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10720056 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21913096 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1354680 - -
          RatRef-12 Expression BeadChip ILMN_66300 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline