Search
Search
| Gene Symbol: Myl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001035252.2 | NP_001030329.2 | 5 - 6 | 385 | 94 | |
| GenBank mRNA | BC126064.1 | - | 5 - 6 | 385 | 7190683 | 94 |
| X07314.1 | - | 5 - 6 | 378 | 94 | ||
Set Membership: |
|