Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01480958_m1
          See other SVOP GE Assays ›
          Gene Symbol:
          Svop
          Gene Name:
          SV2 related protein
          Gene Aliases:
          -
          Chromosome Location:
          Chr.12: 48292284 - 48351974 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01480958_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Svop
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_134404.2 NP_599231.2 15 - 16 1582 59
          XM_008769315.1 XP_008767537.1 15 - 16 1644 59
          XM_008769316.1 XP_008767538.1 15 - 16 2110 59
          GenBank mRNA AF060173.1 - 15 - 16 1739 59

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1200000814.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Transporter 3' Most Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Svop

          Gene Ontology Categories:

          Function(s) Process(es)

          ion transmembrane transport
          transmembrane transport
          ion transmembrane transporter activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P517858 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 889744 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 124498 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 607618 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 479244 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 733479 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 399725 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 207410 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 848511 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 355839 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 952847 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 935421 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 375950 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 221109 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 456706 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 399835 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 568403 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 365427 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 654729 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 744724 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 191371 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 383777 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 532947 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 854045 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 875634 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 857214 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 875427 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 64322 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 400707 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 183879 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 23562 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 714110 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 196180 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 386077 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 440892 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 763215 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 967893 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 333509 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 503290 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 651439 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 896928 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 108516 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 115857 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 159347 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 104381 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 52385 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 157358 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 86913 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 147577 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P517858 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P517858 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P517858 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P517858 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 44434 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 15475 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 101213 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 6940 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 104557 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10759132 - -
          Affymetrix Rat Gene 1.0 ST Array 10759144 - -
          Affymetrix Rat Gene 1.0 ST Array 10759136 - -
          Affymetrix Rat Gene 1.0 ST Array 10759142 - -
          Affymetrix Rat Gene 1.0 ST Array 10759137 - -
          Affymetrix Rat Gene 1.0 ST Array 10759146 - -
          Affymetrix Rat Gene 1.0 ST Array 10759131 - -
          Affymetrix Rat Gene 1.0 ST Array 10759133 - -
          Affymetrix Rat Gene 1.0 ST Array 10759134 - -
          Affymetrix Rat Gene 1.0 ST Array 10759130 - -
          Affymetrix Rat Gene 1.0 ST Array 10759135 - -
          Affymetrix Rat Gene 1.0 ST Array 10759141 - -
          Affymetrix Rat Gene 1.0 ST Array 10759143 - -
          Affymetrix Rat Gene 1.0 ST Array 10759140 - -
          Affymetrix Rat Gene 1.0 ST Array 10759138 - -
          Affymetrix Rat Gene 1.0 ST Array 10759145 - -
          Affymetrix Rat Gene 1.0 ST Array 10759139 - -
          Affymetrix Rat Gene 1.1 ST Array 10759129 - -
          Rat Genome 230 2.0 Array 1369022_at - -
          Rat Genome 230 2.0 Array 1369022_PM_at - -
          Rat Genome 230 2.0 Array 1388845_PM_at - -
          Rat Genome 230 2.0 Array 1388845_at - -
          Affymetrix HT Rat Focus Array 1369022_PM_at - -
          Affymetrix HT Rat Focus Array 1369022_at - -
          Affymetrix Rat Clariom S Assay PSR1200007149.rn.1 TC1200000814.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1200007146.rn.1 TC1200000814.rn.1 RTA TC
          Rat Expression Set 230 1369022_at - -
          Rat Expression Set 230 1369022_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10759129 - -
          Rat Genome U34 Set AF060173_at - -
          Rat Genome U34 Set AF060173_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20727328 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1365477 - -
          RatRef-12 Expression BeadChip ILMN_62205 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline