Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01481057_m1
          See other HPS4 GE Assays ›
          Gene Symbol:
          Hps4
          Gene Name:
          Hermansky-Pudlak syndrome 4
          Gene Aliases:
          -
          Chromosome Location:
          Chr.12: 50285293 - 50315844 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01481057_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 4,430.00
          Your Price
          Online offer:
          4430.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Hps4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001107148.2 NP_001100618.1 10 - 11 1565 90
          XM_006249542.2 XP_006249604.1 10 - 11 2001 90
          XM_006249543.2 XP_006249605.1 10 - 11 1998 90
          XM_006249544.1 XP_006249606.1 11 - 12 2017 90
          XM_006249545.2 XP_006249607.1 10 - 11 2001 90
          XM_006249546.2 XP_006249608.1 10 - 11 2001 90
          XM_008769329.1 XP_008767551.1 11 - 12 2081 90
          GenBank mRNA BC168901.1 - 10 - 11 1565 7385143 90

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1200001825.rn.1

          Set Membership:

          Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          protein targeting
          organelle organization
          lysosome organization
          blood coagulation
          biological_process
          melanocyte differentiation
          protein stabilization
          molecular_function
          protein homodimerization activity
          protein dimerization activity

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Clariom S Assay PSR1200015588.rn.1 TC1200001825.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1200015579.rn.1 TC1200001825.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1200015586.rn.1 TC1200001825.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1200015580.rn.1 TC1200001825.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1200015581.rn.1 TC1200001825.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1200015587.rn.1 TC1200001825.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1200015583.rn.1 TC1200001825.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1200015585.rn.1 TC1200001825.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1200015590.rn.1 TC1200001825.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1200015591.rn.1 TC1200001825.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1200015589.rn.1 TC1200001825.rn.1 RTA TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline