Search
Search
| Gene Symbol: Clip1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031745.2 | NP_113933.2 | 16 - 17 | 3166 | 76 | |
| XM_006249354.2 | XP_006249416.1 | 16 - 17 | 3166 | 76 | ||
| XM_008769250.1 | XP_008767472.1 | 19 - 20 | 5777 | 76 | ||
| XM_008769251.1 | XP_008767473.1 | 18 - 19 | 5738 | 76 | ||
| XM_008769252.1 | XP_008767474.1 | 17 - 18 | 5705 | 76 | ||
| XM_008769253.1 | XP_008767475.1 | 17 - 18 | 5633 | 76 | ||
| XM_008769254.1 | XP_008767476.1 | 16 - 17 | 5600 | 76 | ||
| XM_008769255.1 | XP_008767477.1 | 10 - 11 | 6880 | 76 | ||
| XM_008769256.1 | XP_008767478.1 | 17 - 18 | 3470 | 76 | ||
| XM_008769257.1 | XP_008767479.1 | 17 - 18 | 3470 | 76 | ||
| XM_008769258.1 | XP_008767480.1 | 16 - 17 | 3437 | 76 | ||
| XM_008769259.1 | XP_008767481.1 | 15 - 16 | 3332 | 76 | ||
| GenBank mRNA | AJ237670.1 | - | 16 - 17 | 3166 | 76 | |
Set Membership: |
|