Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01481324_m1
          See other SFSWAP GE Assays ›
          Gene Symbol:
          Sfswap
          Gene Name:
          splicing factor, suppressor of white-apricot family
          Gene Aliases:
          Sfrs8, Srsf8
          Chromosome Location:
          Chr.12: 30740563 - 30811016 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01481324_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Sfswap
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001034924.1 NP_001030096.1 12 - 13 1911 62
          XM_006249269.2 XP_006249331.1 12 - 13 2094 62
          XM_006249270.2 XP_006249332.1 12 - 13 2094 62
          XM_006249271.2 XP_006249333.1 12 - 13 2094 62
          XM_006249273.2 XP_006249335.1 12 - 13 2010 62
          XM_008769196.1 XP_008767418.1 12 - 13 2094 62
          XM_008769197.1 XP_008767419.1 12 - 13 2010 62
          XM_008769198.1 XP_008767420.1 11 - 12 4426 62
          XM_008769199.1 XP_008767421.1 12 - 13 2047 62
          GenBank mRNA BC091384.1 - 8 - 9 1069 62
          BC105868.1 - 12 - 13 1911 7462407 62

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1200001455.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Sfswap

          Panther Classification:

          Molecular Function -

          RNA splicing factor

          Gene Ontology Categories:

          Function(s) Process(es)

          transcription, DNA-templated
          regulation of transcription, DNA-templated
          mRNA processing
          RNA splicing
          negative regulation of mRNA splicing, via spliceosome
          RNA binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P1015606 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 286477 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 173958 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 477736 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 391226 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 296125 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 903657 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 161216 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 356278 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 293319 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 295229 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 299462 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 771784 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 607632 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 521833 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 340005 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 77747 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 928291 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 493077 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 476618 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 160617 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 565797 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 745268 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 351656 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 548075 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 569556 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 76491 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 884846 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 598994 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 644354 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 530107 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 100106 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 487786 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 484316 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 911753 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 416197 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 522802 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 133696 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 663530 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 835635 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 807646 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 18536 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 726380 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 847388 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 611793 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 336646 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 219362 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 318854 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 581318 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 870835 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 823043 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 649599 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 138766 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 166806 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 95262 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 101359 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 69739 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 118778 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 109287 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 178989 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 58390 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P1015606 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P1015606 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P1015606 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P18523 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P1015606 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 44816 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 41073 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 32995 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 70395 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 84933 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 427 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 41714 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10761431 - -
          Affymetrix Rat Gene 1.0 ST Array 10761412 - -
          Affymetrix Rat Gene 1.0 ST Array 10761430 - -
          Affymetrix Rat Gene 1.0 ST Array 10761429 - -
          Affymetrix Rat Gene 1.0 ST Array 10761416 - -
          Affymetrix Rat Gene 1.0 ST Array 10761413 - -
          Affymetrix Rat Gene 1.0 ST Array 10761415 - -
          Affymetrix Rat Gene 1.0 ST Array 10761414 - -
          Affymetrix Rat Gene 1.0 ST Array 10761420 - -
          Affymetrix Rat Gene 1.0 ST Array 10761427 - -
          Affymetrix Rat Gene 1.0 ST Array 10761419 - -
          Affymetrix Rat Gene 1.0 ST Array 10761424 - -
          Affymetrix Rat Gene 1.0 ST Array 10761423 - -
          Affymetrix Rat Gene 1.0 ST Array 10761433 - -
          Affymetrix Rat Gene 1.0 ST Array 10761425 - -
          Affymetrix Rat Gene 1.0 ST Array 10761418 - -
          Affymetrix Rat Gene 1.0 ST Array 10761434 - -
          Affymetrix Rat Gene 1.0 ST Array 10761426 - -
          Affymetrix Rat Gene 1.0 ST Array 10761428 - -
          Affymetrix Rat Gene 1.1 ST Array 10761411 - -
          Rat Genome 230 2.0 Array 1390919_at - -
          Rat Genome 230 2.0 Array 1383676_at - -
          Rat Genome 230 2.0 Array 1383676_PM_at - -
          Rat Genome 230 2.0 Array 1390919_PM_at - -
          Rat Genome 230 2.0 Array 1391858_at - -
          Rat Genome 230 2.0 Array 1391858_PM_at - -
          Affymetrix HT Rat Focus Array 1391858_PM_at - -
          Affymetrix HT Rat Focus Array 1383676_PM_at - -
          Affymetrix HT Rat Focus Array 1391858_at - -
          Affymetrix HT Rat Focus Array 1383676_at - -
          Affymetrix Rat Clariom S Assay PSR1200012300.rn.1 TC1200001455.rn.1 RTA TC
          Rat Expression Set 230 1391858_at - -
          Rat Expression Set 230 1383676_PM_at - -
          Rat Expression Set 230 1383676_at - -
          Rat Expression Set 230 1391858_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10761411 - -
          Rat Genome U34 Set rc_AI043919_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21877441 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1650597 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline