Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01481341_m1
          See other SERPINE1 GE Assays ›
          Gene Symbol:
          Serpine1
          Gene Name:
          serpin peptidase inhibitor, clade E (nexin, plasminogen activator inhibitor type 1), member 1
          Gene Aliases:
          PAI1A, Pai1, Pai1aa, Planh, RATPAI1A
          Chromosome Location:
          Chr.12: 22641104 - 22651482 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01481341_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Serpine1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_012620.1 NP_036752.1 7 - 8 1205 59
          GenBank mRNA M24067.1 - 7 - 8 1205 59

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1200000390.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Recommended Assay Rattus norvegicus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Serpine1

          Panther Classification:

          Molecular Function -

          protease inhibitor

          Gene Ontology Categories:

          Function(s) Process(es)

          response to reactive oxygen species
          chronological cell aging
          angiogenesis
          female gonad development
          regulation of receptor activity
          positive regulation of gene expression
          negative regulation of gene expression
          negative regulation of plasminogen activation
          negative regulation of endopeptidase activity
          negative regulation of smooth muscle cell migration
          positive regulation of blood coagulation
          negative regulation of cell migration
          response to lipopolysaccharide
          positive regulation of interleukin-8 production
          cellular response to insulin stimulus
          negative regulation of cell adhesion mediated by integrin
          response to cytokine
          response to laminar fluid shear stress
          positive regulation of leukotriene production involved in inflammatory response
          cellular response to drug
          cellular response to potassium ion
          cellular response to platelet-derived growth factor stimulus
          wound healing
          regulation of cell proliferation
          tissue regeneration
          glucose homeostasis
          response to starvation
          response to estrogen
          cellular response to fibroblast growth factor stimulus
          mast cell activation
          regulation of angiogenesis
          positive regulation of angiogenesis
          negative regulation of proteolysis
          decidualization
          positive regulation of fibroblast proliferation
          positive regulation of receptor-mediated endocytosis
          positive regulation of inflammatory response
          positive regulation of coagulation
          defense response to Gram-negative bacterium
          positive regulation of keratinocyte migration
          negative regulation of fibrinolysis
          response to hyperoxia
          negative regulation of vascular wound healing
          cellular response to hydrogen peroxide
          response to epidermal growth factor
          cellular response to lipopolysaccharide
          cellular response to metal ion
          cellular response to gravity
          cellular response to mechanical stimulus
          cellular response to nicotine
          cellular response to ATP
          cellular response to cAMP
          cellular response to cGMP
          cellular response to glucose stimulus
          cellular response to interleukin-1
          cellular response to growth factor stimulus
          cellular response to follicle-stimulating hormone stimulus
          cellular response to glucagon stimulus
          cellular response to low-density lipoprotein particle stimulus
          cellular response to organic cyclic compound
          cellular response to cycloheximide
          cellular response to hypoxia
          cellular response to dexamethasone stimulus
          response to transforming growth factor beta
          cellular response to transforming growth factor beta stimulus
          positive regulation of monocyte chemotaxis
          positive regulation of calcium ion import
          negative regulation of extrinsic apoptotic signaling pathway via death domain receptors
          cellular response to nerve growth factor stimulus
          negative regulation of smooth muscle cell-matrix adhesion
          negative regulation of endothelial cell apoptotic process
          protease binding
          serine-type endopeptidase inhibitor activity
          receptor binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Clariom S Assay PSR1200003374.rn.1 TC1200000390.rn.1 RTA TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-h9fps:80/100.66.77.141:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline