Search
Search
| Gene Symbol: Zcchc8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001105929.1 | NP_001099399.1 | 1 - 2 | 295 | 79 | |
| XM_006249333.2 | XP_006249395.1 | 1 - 2 | 390 | 79 | ||
| XM_006249334.2 | XP_006249396.1 | 1 - 2 | 390 | 79 | ||
Set Membership: |
|