Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01481566_m1
          See other RAD9B GE Assays ›
          Gene Symbol:
          Rad9b
          Gene Name:
          RAD9 homolog B (S. pombe)
          Gene Aliases:
          -
          Chromosome Location:
          Chr.12: 39699140 - 39731218 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01481566_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rad9b
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001030042.1 NP_001025213.1 10 - 11 1245 80
          XM_006249366.2 XP_006249428.1 10 - 11 1871 80
          XM_008769241.1 XP_008767463.1 9 - 10 1139 80
          XM_008769242.1 XP_008767464.1 10 - 11 1263 80
          XM_008769243.1 XP_008767465.1 9 - 10 1064 80
          XM_008769244.1 XP_008767466.1 10 - 11 1159 80
          XM_008769245.1 XP_008767467.1 10 - 11 1098 80
          XM_008769246.1 XP_008767468.1 10 - 11 1092 80
          GenBank mRNA BC099750.1 - 10 - 11 1245 5599441 80

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1200000651.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Rad9b

          Panther Classification:

          Molecular Function -

          exodeoxyribonuclease

          Gene Ontology Categories:

          Function(s)

          DNA damage checkpoint
          DNA repair

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P031219 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 386304 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 424594 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 673541 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 677248 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 249186 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 311694 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 131625 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 805213 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 372837 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 149792 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 358820 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 328944 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 713254 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 157901 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 361089 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 743157 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 191594 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 953973 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 211617 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 544604 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 23156 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 57386 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 143351 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 905435 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 522283 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 178725 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 137510 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 146792 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 94741 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 146623 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P031219 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P031219 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P031219 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P363330 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 58650 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 77655 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 3378 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 76929 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10758594 - -
          Affymetrix Rat Gene 1.0 ST Array 10758589 - -
          Affymetrix Rat Gene 1.0 ST Array 10758597 - -
          Affymetrix Rat Gene 1.0 ST Array 10758598 - -
          Affymetrix Rat Gene 1.0 ST Array 10758590 - -
          Affymetrix Rat Gene 1.0 ST Array 10758588 - -
          Affymetrix Rat Gene 1.0 ST Array 10758592 - -
          Affymetrix Rat Gene 1.0 ST Array 10758595 - -
          Affymetrix Rat Gene 1.0 ST Array 10758596 - -
          Affymetrix Rat Gene 1.0 ST Array 10758591 - -
          Affymetrix Rat Gene 1.0 ST Array 10758593 - -
          Affymetrix Rat Gene 1.1 ST Array 10758587 - -
          Rat Genome 230 2.0 Array 1396511_PM_at - -
          Rat Genome 230 2.0 Array 1384876_PM_at - -
          Rat Genome 230 2.0 Array 1396511_at - -
          Rat Genome 230 2.0 Array 1384876_at - -
          Affymetrix HT Rat Focus Array 1384876_PM_at - -
          Affymetrix HT Rat Focus Array 1384876_at - -
          Affymetrix Rat Clariom S Assay PSR1200005792.rn.1 TC1200000651.rn.1 RTA TC
          Rat Expression Set 230 1384876_PM_at - -
          Rat Expression Set 230 1384876_at - -
          Affymetrix Rat Gene 1.0 ST Array 10758587 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21081793 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1364902 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline