Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01481627_m1
          See other POLE GE Assays ›
          Gene Symbol:
          Pole
          Gene Name:
          polymerase (DNA directed), epsilon, catalytic subunit
          Gene Aliases:
          -
          Chromosome Location:
          Chr.12: 52403533 - 52452079 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01481627_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Pole
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001107152.2 NP_001100622.2 23 - 24 2746 138
          XM_006249562.2 XP_006249624.1 23 - 24 2783 138
          XM_008769335.1 XP_008767557.1 23 - 24 2783 138

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1200001869.rn.1

          Set Membership:

          Amplicon length greater than or equal to 101 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Panther Classification:

          Molecular Function -

          DNA metabolism protein

          Gene Ontology Categories:

          Function(s) Process(es)

          G1/S transition of mitotic cell cycle
          DNA synthesis involved in DNA repair
          in utero embryonic development
          DNA replication
          base-excision repair, gap-filling
          nucleotide-excision repair, DNA gap filling
          DNA biosynthetic process
          nucleotide binding
          molecular_function
          DNA binding
          chromatin binding
          DNA-directed DNA polymerase activity
          zinc ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P431150 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 440729 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 927192 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 446614 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 912152 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 759905 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 938943 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 568843 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 809291 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 918489 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 911967 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 346001 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 63185 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 781493 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 327124 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 47471 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 734547 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 956465 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 67102 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 365403 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 943639 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 908649 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 815552 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 95403 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 918238 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 516320 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 381629 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 528904 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 464337 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 694900 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 756449 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 584725 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 661170 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 524480 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 24956 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 141459 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 583934 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 24832 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 59083 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 157034 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 40823 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 35820 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 14673 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P431150 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P431150 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P431150 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P431150 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 104161 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 64946 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 53401 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 15954 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10763035 - -
          Affymetrix Rat Gene 1.0 ST Array 10763027 - -
          Affymetrix Rat Gene 1.0 ST Array 10763022 - -
          Affymetrix Rat Gene 1.0 ST Array 10763067 - -
          Affymetrix Rat Gene 1.0 ST Array 10763041 - -
          Affymetrix Rat Gene 1.0 ST Array 10763039 - -
          Affymetrix Rat Gene 1.0 ST Array 10763062 - -
          Affymetrix Rat Gene 1.0 ST Array 10763033 - -
          Affymetrix Rat Gene 1.0 ST Array 10763037 - -
          Affymetrix Rat Gene 1.0 ST Array 10763030 - -
          Affymetrix Rat Gene 1.0 ST Array 10763048 - -
          Affymetrix Rat Gene 1.0 ST Array 10763051 - -
          Affymetrix Rat Gene 1.0 ST Array 10763034 - -
          Affymetrix Rat Gene 1.0 ST Array 10763040 - -
          Affymetrix Rat Gene 1.0 ST Array 10763054 - -
          Affymetrix Rat Gene 1.0 ST Array 10763070 - -
          Affymetrix Rat Gene 1.0 ST Array 10763057 - -
          Affymetrix Rat Gene 1.0 ST Array 10763024 - -
          Affymetrix Rat Gene 1.0 ST Array 10763045 - -
          Affymetrix Rat Gene 1.0 ST Array 10763043 - -
          Affymetrix Rat Gene 1.0 ST Array 10763047 - -
          Affymetrix Rat Gene 1.0 ST Array 10763072 - -
          Affymetrix Rat Gene 1.0 ST Array 10763064 - -
          Affymetrix Rat Gene 1.0 ST Array 10763058 - -
          Affymetrix Rat Gene 1.0 ST Array 10763046 - -
          Affymetrix Rat Gene 1.0 ST Array 10763050 - -
          Affymetrix Rat Gene 1.0 ST Array 10763028 - -
          Affymetrix Rat Gene 1.0 ST Array 10763066 - -
          Affymetrix Rat Gene 1.0 ST Array 10763061 - -
          Affymetrix Rat Gene 1.0 ST Array 10763042 - -
          Affymetrix Rat Gene 1.0 ST Array 10763056 - -
          Affymetrix Rat Gene 1.0 ST Array 10763023 - -
          Affymetrix Rat Gene 1.0 ST Array 10763068 - -
          Affymetrix Rat Gene 1.0 ST Array 10763032 - -
          Affymetrix Rat Gene 1.0 ST Array 10763036 - -
          Affymetrix Rat Gene 1.0 ST Array 10763059 - -
          Affymetrix Rat Gene 1.0 ST Array 10763055 - -
          Affymetrix Rat Gene 1.0 ST Array 10763053 - -
          Affymetrix Rat Gene 1.0 ST Array 10763049 - -
          Affymetrix Rat Gene 1.0 ST Array 10763052 - -
          Affymetrix Rat Gene 1.0 ST Array 10763044 - -
          Affymetrix Rat Gene 1.0 ST Array 10763060 - -
          Affymetrix Rat Gene 1.0 ST Array 10763071 - -
          Affymetrix Rat Gene 1.0 ST Array 10763069 - -
          Affymetrix Rat Gene 1.0 ST Array 10763063 - -
          Affymetrix Rat Gene 1.0 ST Array 10763038 - -
          Affymetrix Rat Gene 1.0 ST Array 10763025 - -
          Affymetrix Rat Gene 1.0 ST Array 10763065 - -
          Affymetrix Rat Gene 1.0 ST Array 10763026 - -
          Affymetrix Rat Gene 1.1 ST Array 10763021 - -
          Rat Genome 230 2.0 Array 1389421_at - -
          Rat Genome 230 2.0 Array 1389421_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1200015886.rn.1 TC1200001869.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1200015887.rn.1 TC1200001869.rn.1 RTA TC
          Rat Expression Set 230 1389421_at - -
          Rat Expression Set 230 1389421_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10763021 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22116377 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1366864 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0