Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01481654_m1
          See other TFR2 GE Assays ›
          Gene Symbol:
          Tfr2
          Gene Name:
          transferrin receptor 2
          Gene Aliases:
          Trfr2
          Chromosome Location:
          Chr.12: 22177382 - 22194333 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01481654_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tfr2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001105916.1 NP_001099386.1 11 - 12 1441 66
          XM_006249116.2 XP_006249178.1 10 - 11 1393 66
          XM_006249117.1 XP_006249179.1 11 - 12 1409 66
          XM_006249118.1 XP_006249180.1 11 - 12 1412 66
          XM_008769125.1 XP_008767347.1 10 - 11 1522 66
          XM_008769126.1 XP_008767348.1 10 - 11 1352 66
          XM_008769127.1 XP_008767349.1 10 - 11 1393 66
          XM_008769128.1 XP_008767350.1 10 - 11 1393 66
          GenBank mRNA BC166451.1 - 10 - 11 1506 8371649 66

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1200001298.rn.1

          Set Membership:

          Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Panther Classification:

          Molecular Function -

          metalloprotease

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          iron ion homeostasis
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P22470 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 119657 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 678225 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 640756 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 11553 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 642400 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 625007 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 295786 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 457034 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 514580 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 283291 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 891717 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 37229 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 917538 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 180749 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 80247 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 129977 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P22470 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P22470 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P22470 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P22470 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 46567 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 50952 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10757375 - -
          Affymetrix Rat Gene 1.0 ST Array 10757386 - -
          Affymetrix Rat Gene 1.0 ST Array 10757380 - -
          Affymetrix Rat Gene 1.0 ST Array 10757373 - -
          Affymetrix Rat Gene 1.0 ST Array 10757389 - -
          Affymetrix Rat Gene 1.0 ST Array 10757382 - -
          Affymetrix Rat Gene 1.0 ST Array 10757385 - -
          Affymetrix Rat Gene 1.0 ST Array 10757379 - -
          Affymetrix Rat Gene 1.0 ST Array 10757383 - -
          Affymetrix Rat Gene 1.0 ST Array 10757388 - -
          Affymetrix Rat Gene 1.0 ST Array 10757376 - -
          Affymetrix Rat Gene 1.0 ST Array 10757377 - -
          Affymetrix Rat Gene 1.0 ST Array 10757387 - -
          Affymetrix Rat Gene 1.0 ST Array 10757381 - -
          Affymetrix Rat Gene 1.0 ST Array 10757378 - -
          Affymetrix Rat Gene 1.0 ST Array 10757374 - -
          Affymetrix Rat Gene 1.0 ST Array 10757384 - -
          Affymetrix Rat Gene 1.1 ST Array 10757372 - -
          Rat Genome 230 2.0 Array 1380066_at - -
          Rat Genome 230 2.0 Array 1380066_PM_at - -
          Affymetrix HT Rat Focus Array 1380066_at - -
          Affymetrix HT Rat Focus Array 1380066_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1200011130.rn.1 TC1200001298.rn.1 RTA TC
          Rat Expression Set 230 1380066_PM_at - -
          Rat Expression Set 230 1380066_at - -
          Affymetrix Rat Gene 1.0 ST Array 10757372 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21030436 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_2038910 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline