Search Thermo Fisher Scientific
- Contáctenos
- Orden Rápida
-
¿No tiene una cuenta? Crear una cuenta
Search Thermo Fisher Scientific
Gene Symbol: Mrps17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105923.1 | NP_001099393.1 | 2 - 3 | 481 | 80 | |
XM_006249264.2 | XP_006249326.1 | 2 - 3 | 201 | 80 | ||
GenBank mRNA | FQ223326.1 | - | 2 - 3 | 155 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons |