Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01481975_m1
          See other REC8 GE Assays ›
          Gene Symbol:
          Rec8
          Gene Name:
          REC8 meiotic recombination protein
          Gene Aliases:
          Rec8L1
          Chromosome Location:
          Chr.15: 34296922 - 34304387 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01481975_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rec8
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001011916.1 NP_001011916.1 7 - 8 761 116
          GenBank mRNA AY529701.1 - 7 - 8 777 116
          BC079023.1 - 7 - 8 761 7113870 116

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1500000515.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Rattus norvegicus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Rec8

          Gene Ontology Categories:

          Function(s) Process(es)

          double-strand break repair via homologous recombination
          oocyte maturation
          meiotic nuclear division
          synapsis
          male meiosis I
          spermatid development
          fertilization
          linear element assembly
          seminiferous tubule development
          protein binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P025639 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 592280 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 62303 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 646423 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 110441 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 874085 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 317372 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P025639 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P025639 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P025639 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P320812 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 46835 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10780463 - -
          Affymetrix Rat Gene 1.0 ST Array 10780452 - -
          Affymetrix Rat Gene 1.0 ST Array 10780450 - -
          Affymetrix Rat Gene 1.0 ST Array 10780451 - -
          Affymetrix Rat Gene 1.0 ST Array 10780454 - -
          Affymetrix Rat Gene 1.0 ST Array 10780459 - -
          Affymetrix Rat Gene 1.0 ST Array 10780465 - -
          Affymetrix Rat Gene 1.0 ST Array 10780462 - -
          Affymetrix Rat Gene 1.0 ST Array 10780456 - -
          Affymetrix Rat Gene 1.0 ST Array 10780453 - -
          Affymetrix Rat Gene 1.0 ST Array 10780466 - -
          Affymetrix Rat Gene 1.0 ST Array 10780449 - -
          Affymetrix Rat Gene 1.0 ST Array 10780458 - -
          Affymetrix Rat Gene 1.0 ST Array 10780460 - -
          Affymetrix Rat Gene 1.0 ST Array 10780447 - -
          Affymetrix Rat Gene 1.0 ST Array 10780457 - -
          Affymetrix Rat Gene 1.0 ST Array 10780467 - -
          Affymetrix Rat Gene 1.0 ST Array 10780464 - -
          Affymetrix Rat Gene 1.0 ST Array 10780448 - -
          Affymetrix Rat Gene 1.0 ST Array 10780461 - -
          Affymetrix Rat Gene 1.0 ST Array 10780455 - -
          Affymetrix Rat Gene 1.1 ST Array 10780446 - -
          Affymetrix Rat Clariom S Assay PSR1500003468.rn.1 TC1500000515.rn.1 RTA TC
          Affymetrix Rat Gene 1.0 ST Array 10780446 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1376718 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline