Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Tubes
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO 和 CRO 服務
    • CDMO 服務
    • 臨床試驗 CRO 服務
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Register for an Account
    • How to Order
    • Instrument Support
    • Technical Support Centers
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • 快速訂購
          • 登入
            登入
            Don't have an account ? Create Account
            • 帳戶
            • 訂單
            • 訂製產品與專案
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01482017_m1
          See other LOC290071 GE Assays ›
          Gene Symbol:
          LOC290071
          Gene Name:
          similar to RIKEN cDNA A430107P09 gene
          Gene Aliases:
          RGD1359684, Tcra, Tra29
          Chromosome Location:
          -
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01482017_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 4,430.00
          Your Price
          Online offer:
          4430.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LOC290071
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA M18853.1 - 3 - 4 382 77

          Back To Top

          More Information


          Set Membership:

          Amplicon length between 71 and 85 3' Most Recommended Assay Rattus norvegicus Probe spans exons

          Panther Classification:

          Molecular Function -

          immunoglobulin receptor superfamily

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 883673 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 682733 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 587085 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 935686 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 418355 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 299500 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 698673 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 473937 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 56733 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 600724 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 964929 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 74768 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 828147 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 526977 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 454304 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 527565 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 953456 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 325664 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 680874 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 111386 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 545553 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 409268 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 14840 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 53687 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 74873 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 133428 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 67977 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 7325 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 61213 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 179112 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 80029 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 17891 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 155140 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P213045 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P405374 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 18619 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 75157 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 21776 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 56964 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 33635 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 18054 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 84399 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 92893 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 89627 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10783508 - -
          Affymetrix Rat Gene 1.0 ST Array 10780069 - -
          Affymetrix Rat Gene 1.0 ST Array 10780027 - -
          Affymetrix Rat Gene 1.0 ST Array 10780125 - -
          Affymetrix Rat Gene 1.0 ST Array 10780123 - -
          Affymetrix Rat Gene 1.0 ST Array 10780022 - -
          Affymetrix Rat Gene 1.0 ST Array 10783379 - -
          Affymetrix Rat Gene 1.0 ST Array 10780118 - -
          Affymetrix Rat Gene 1.0 ST Array 10780122 - -
          Affymetrix Rat Gene 1.0 ST Array 10780126 - -
          Affymetrix Rat Gene 1.0 ST Array 10783380 - -
          Affymetrix Rat Gene 1.0 ST Array 10779993 - -
          Affymetrix Rat Gene 1.0 ST Array 10783407 - -
          Affymetrix Rat Gene 1.0 ST Array 10780068 - -
          Affymetrix Rat Gene 1.0 ST Array 10783423 - -
          Affymetrix Rat Gene 1.0 ST Array 10780124 - -
          Affymetrix Rat Gene 1.0 ST Array 10783424 - -
          Affymetrix Rat Gene 1.0 ST Array 10780121 - -
          Affymetrix Rat Gene 1.0 ST Array 10780028 - -
          Affymetrix Rat Gene 1.1 ST Array 10780026 - -
          Affymetrix Rat Gene 1.1 ST Array 10780020 - -
          Affymetrix Rat Gene 1.1 ST Array 10780094 - -
          Affymetrix Rat Gene 1.1 ST Array 10783378 - -
          Affymetrix Rat Gene 1.1 ST Array 10783419 - -
          Rat Genome 230 2.0 Array 1371016_PM_at - -
          Rat Genome 230 2.0 Array 1371016_at - -
          Rat Expression Set 230 1371016_at - -
          Rat Expression Set 230 1371016_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10779990 - -
          Affymetrix Rat Gene 1.0 ST Array 10780020 - -
          Affymetrix Rat Gene 1.0 ST Array 10783419 - -
          Affymetrix Rat Gene 1.0 ST Array 10783406 - -
          Affymetrix Rat Gene 1.0 ST Array 10780065 - -
          Affymetrix Rat Gene 1.0 ST Array 10780094 - -
          Affymetrix Rat Gene 1.0 ST Array 10783378 - -
          Rat Genome U34 Set L11035_g_at - -
          Rat Genome U34 Set X14318cds_at - -
          Rat Genome U34 Set X62327cds_r_at - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1650986 - -
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1354082 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline