Search
Search
| Gene Symbol: Carkd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108402.1 | NP_001101872.1 | 2 - 3 | 221 | 80 | |
| XM_006253465.2 | XP_006253527.1 | 2 - 3 | 348 | 80 | ||
| XM_008771403.1 | XP_008769625.1 | 2 - 3 | 239 | 80 | ||
| GenBank mRNA | FQ225763.1 | - | 183 | 80 | ||
Set Membership: |
|