Search
Search
| Gene Symbol: Thsd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108878.1 | NP_001102348.1 | 2 - 3 | 264 | 69 | |
| XM_006253357.2 | XP_006253419.1 | 2 - 3 | 1682 | 69 | ||
| GenBank mRNA | BC162035.1 | - | 1 - 2 | 135 | 7097232 | 69 |
Set Membership: |
|