Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01482295_m1
          See other GAS6 GE Assays ›
          Gene Symbol:
          Gas6
          Gene Name:
          growth arrest specific 6
          Gene Aliases:
          -
          Chromosome Location:
          Chr.16: 81213364 - 81243757 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01482295_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gas6
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_057100.2 NP_476441.2 14 - 15 2000 58
          GenBank mRNA BC070881.1 - 14 - 15 2000 7096665 58
          D42148.1 - 14 - 15 1895 58

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1600002008.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Gas6

          Panther Classification:

          Molecular Function -

          extracellular matrix protein

          Gene Ontology Categories:

          Function(s) Process(es)

          neuron migration
          positive regulation of protein phosphorylation
          positive regulation of cytokine-mediated signaling pathway
          positive regulation of glomerular filtration
          protein phosphorylation
          phagocytosis
          signal transduction
          enzyme linked receptor protein signaling pathway
          blood coagulation
          cellular response to starvation
          positive regulation of gene expression
          negative regulation of tumor necrosis factor-mediated signaling pathway
          macrophage cytokine production
          peptidyl-serine phosphorylation
          fusion of virus membrane with host plasma membrane
          viral genome replication
          organ regeneration
          cell-substrate adhesion
          positive regulation of TOR signaling
          activation of protein kinase B activity
          negative regulation of interferon-gamma production
          negative regulation of interleukin-6 production
          negative regulation of tumor necrosis factor production
          positive regulation of natural killer cell differentiation
          positive regulation of peptidyl-serine phosphorylation
          negative regulation of protein import into nucleus, translocation
          cellular response to interferon-alpha
          cellular response to drug
          B cell chemotaxis
          regulation of growth
          tissue regeneration
          negative regulation of apoptotic process
          negative regulation of cysteine-type endopeptidase activity involved in apoptotic process
          apoptotic cell clearance
          negative regulation of sequence-specific DNA binding transcription factor activity
          protein kinase B signaling
          positive regulation of protein kinase activity
          negative regulation of transcription, DNA-templated
          viral entry into host cell
          receptor-mediated virion attachment to host cell
          positive regulation of protein export from nucleus
          positive regulation of fibroblast proliferation
          tissue remodeling
          negative regulation of interleukin-1 secretion
          positive regulation of phagocytosis
          positive regulation of protein kinase B signaling
          positive regulation of protein tyrosine kinase activity
          negative regulation of biomineral tissue development
          positive regulation of ERK1 and ERK2 cascade
          calcium ion transmembrane transport
          cellular response to vitamin K
          cellular response to glucose stimulus
          cellular response to growth factor stimulus
          protein targeting to plasma membrane
          extracellular matrix assembly
          dendritic cell differentiation
          hematopoietic stem cell migration to bone marrow
          negative regulation of oligodendrocyte apoptotic process
          negative regulation of interleukin-6 secretion
          negative regulation of fibroblast apoptotic process
          positive regulation of receptor activity
          negative regulation of endothelial cell apoptotic process
          positive regulation of dendritic cell chemotaxis
          negative regulation of renal albumin absorption
          negative regulation of dendritic cell apoptotic process
          phosphatidylserine binding
          receptor binding
          voltage-gated calcium channel activity
          calcium ion binding
          growth factor activity
          protein tyrosine kinase activator activity
          receptor tyrosine kinase binding
          cysteine-type endopeptidase inhibitor activity involved in apoptotic process
          receptor agonist activity
          binding, bridging

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P179814 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 28332 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 569984 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 725647 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 849972 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 194616 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 694774 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 9397 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 889214 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 567282 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 805836 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 9727 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 479796 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 887568 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 613170 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 264645 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 552113 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 188237 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 263060 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 22045 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 233289 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 84823 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 114762 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 52802 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P179814 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P179814 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P179814 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P179811 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P179814 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P13025 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 18041 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 8237 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 3154 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 42888 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 9642 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10789453 - -
          Affymetrix Rat Gene 1.0 ST Array 10789456 - -
          Affymetrix Rat Gene 1.0 ST Array 10789454 - -
          Affymetrix Rat Gene 1.0 ST Array 10789455 - -
          Affymetrix Rat Gene 1.0 ST Array 10789451 - -
          Affymetrix Rat Gene 1.0 ST Array 10789457 - -
          Affymetrix Rat Gene 1.0 ST Array 10789445 - -
          Affymetrix Rat Gene 1.0 ST Array 10789459 - -
          Affymetrix Rat Gene 1.0 ST Array 10789446 - -
          Affymetrix Rat Gene 1.0 ST Array 10789444 - -
          Affymetrix Rat Gene 1.0 ST Array 10789448 - -
          Affymetrix Rat Gene 1.0 ST Array 10789452 - -
          Affymetrix Rat Gene 1.0 ST Array 10789447 - -
          Affymetrix Rat Gene 1.0 ST Array 10789458 - -
          Affymetrix Rat Gene 1.0 ST Array 10789449 - -
          Affymetrix Rat Gene 1.0 ST Array 10789443 - -
          Affymetrix Rat Gene 1.0 ST Array 10789450 - -
          Affymetrix Rat Gene 1.1 ST Array 10789442 - -
          Rat Genome 230 2.0 Array 1392784_PM_at - -
          Rat Genome 230 2.0 Array 1383047_at - -
          Rat Genome 230 2.0 Array 1369735_at - -
          Rat Genome 230 2.0 Array 1369735_PM_at - -
          Rat Genome 230 2.0 Array 1392784_at - -
          Rat Genome 230 2.0 Array 1383047_PM_at - -
          Affymetrix HT Rat Focus Array 1369735_at - -
          Affymetrix HT Rat Focus Array 1392784_PM_at - -
          Affymetrix HT Rat Focus Array 1392784_at - -
          Affymetrix HT Rat Focus Array 1369735_PM_at - -
          Affymetrix HT Rat Focus Array 1383047_at - -
          Affymetrix HT Rat Focus Array 1383047_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1600014813.rn.1 TC1600002008.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1600014814.rn.1 TC1600002008.rn.1 RTA TC
          Rat Expression Set 230 1392784_PM_at - -
          Rat Expression Set 230 1369735_at - -
          Rat Expression Set 230 1392784_at - -
          Rat Expression Set 230 1369735_PM_at - -
          Rat Expression Set 230 1383047_at - -
          Rat Expression Set 230 1383047_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10789442 - -
          Rat Genome U34 Set rc_AA874830_at - -
          Rat Genome U34 Set D42148_at - -
          Rat Genome U34 Set rc_AA957335_at - -
          Rat Genome U34 Set rc_AI229260_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21693183 - -
          Array Name Probe IDs Targets Target Type
          RatRef-12 Expression BeadChip ILMN_58726 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline