Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01482578_m1
          See other PLAT GE Assays ›
          Gene Symbol:
          Plat
          Gene Name:
          plasminogen activator, tissue
          Gene Aliases:
          PATISS, tPA
          Chromosome Location:
          Chr.16: 74098263 - 74122897 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01482578_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Plat
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_013151.2 NP_037283.2 13 - 14 1625 77
          XM_006253347.2 XP_006253409.1 13 - 14 1697 77
          XM_008771348.1 XP_008769570.1 14 - 15 1755 77
          GenBank mRNA BC061565.1 - 13 - 14 1625 6920438 77
          M23697.1 - 12 - 13 1549 77

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1600001909.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Protease 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Plat

          Panther Classification:

          Molecular Function -

          serine protease

          Gene Ontology Categories:

          Function(s) Process(es)

          response to hypoxia
          proteolysis
          smooth muscle cell migration
          plasminogen activation
          synaptic transmission, glutamatergic
          wound healing
          response to peptide hormone
          negative regulation of proteolysis
          platelet-derived growth factor receptor signaling pathway
          regulation of synaptic plasticity
          response to cAMP
          positive regulation of ovulation
          cellular response to dexamethasone stimulus
          molecular_function
          serine-type endopeptidase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P119633 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 326214 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 415938 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 89502 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 601471 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 674607 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 139271 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 34907 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 46651 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 535266 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 110152 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 623719 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 443325 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 291829 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 743536 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 320318 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 909130 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 70317 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 89251 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 67800 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 67458 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 70384 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P119633 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P119633 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P119633 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P432432 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700528745 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 46208 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 2471 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 31166 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 15157 - -
          Agilent Rat cDNA Microarray (G4105A) 700528745 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10792430 - -
          Affymetrix Rat Gene 1.0 ST Array 10792423 - -
          Affymetrix Rat Gene 1.0 ST Array 10792435 - -
          Affymetrix Rat Gene 1.0 ST Array 10792431 - -
          Affymetrix Rat Gene 1.0 ST Array 10792429 - -
          Affymetrix Rat Gene 1.0 ST Array 10792422 - -
          Affymetrix Rat Gene 1.0 ST Array 10792434 - -
          Affymetrix Rat Gene 1.0 ST Array 10792432 - -
          Affymetrix Rat Gene 1.0 ST Array 10792427 - -
          Affymetrix Rat Gene 1.0 ST Array 10792424 - -
          Affymetrix Rat Gene 1.0 ST Array 10792428 - -
          Affymetrix Rat Gene 1.0 ST Array 10792433 - -
          Affymetrix Rat Gene 1.0 ST Array 10792426 - -
          Affymetrix Rat Gene 1.0 ST Array 10792425 - -
          Affymetrix Rat Gene 1.1 ST Array 10792421 - -
          Rat Genome 230 2.0 Array 1367800_at - -
          Rat Genome 230 2.0 Array 1367800_PM_at - -
          Affymetrix HT Rat Focus Array 1367800_PM_at - -
          Affymetrix HT Rat Focus Array 1367800_at - -
          Affymetrix Rat Clariom S Assay PSR1600014120.rn.1 TC1600001909.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1600014121.rn.1 TC1600001909.rn.1 RTA TC
          Rat Expression Set 230 1367800_PM_at - -
          Rat Expression Set 230 1367800_at - -
          Affymetrix Rat Gene 1.0 ST Array 10792421 - -
          Rat Genome U34 Set M23697_at - -
          Rat Genome U34 Set M23697_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21634180 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1358127 - -
          RatRef-12 Expression BeadChip ILMN_66750 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline