Search
Search
| Gene Symbol: Adam3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_020302.1 | NP_064698.1 | 18 - 19 | 2131 | 66 | |
| GenBank mRNA | BC078839.1 | - | 18 - 19 | 2037 | 7114315 | 66 |
| Y07903.1 | - | 18 - 19 | 2131 | 66 | ||
Set Membership: |
|