Search
Search
| Gene Symbol: Glul | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_017073.3 | NP_058769.4 | 3 - 4 | 450 | 89 | |
| GenBank mRNA | BC061559.1 | - | 3 - 4 | 463 | 6918514 | 89 |
| BC072694.1 | - | 3 - 4 | 450 | 7103110 | 89 | |
| FQ216093.1 | - | 3 - 4 | 455 | 89 | ||
| FQ226911.1 | - | 3 - 4 | 473 | 89 | ||
| FQ227474.1 | - | 3 - 4 | 462 | 89 | ||
| FQ232853.1 | - | 3 - 4 | 511 | 89 | ||
| FQ232963.1 | - | 3 - 4 | 462 | 89 | ||
| FQ233770.1 | - | 3 - 4 | 462 | 89 | ||
| FQ234289.1 | - | 3 - 4 | 463 | 89 | ||
| M29579.1 | - | 3 - 4 | 461 | 89 | ||
| M91652.1 | - | 3 - 4 | 459 | 89 | ||
| X07921.1 | - | 3 - 4 | 461 | 89 | ||
Set Membership: |
|