Search
Search
| Gene Symbol: Atp2b4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005871.1 | NP_001005871.1 | 14 - 15 | 2567 | 67 | |
| XM_008769446.1 | XP_008767668.1 | 15 - 16 | 2950 | 67 | ||
| XM_008769447.1 | XP_008767669.1 | 15 - 16 | 3587 | 67 | ||
| XM_008769448.1 | XP_008767670.1 | 14 - 15 | 2914 | 67 | ||
| XM_008769449.1 | XP_008767671.1 | 15 - 16 | 2951 | 67 | ||
| XM_008769450.1 | XP_008767672.1 | 14 - 15 | 2915 | 67 | ||
| GenBank mRNA | U15408.1 | - | 14 - 15 | 2567 | 67 | |
Set Membership: |
|