Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01483459_m1
          See other RCSD1 GE Assays ›
          Gene Symbol:
          Rcsd1
          Gene Name:
          RCSD domain containing 1
          Gene Aliases:
          RGD1304715
          Chromosome Location:
          Chr.13: 83854650 - 83917037 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01483459_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 4,430.00
          Your Price
          Online offer:
          4430.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rcsd1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001108349.1 NP_001101819.1 4 - 5 562 68
          XM_006250181.2 XP_006250243.1 5 - 6 688 68
          XM_008769663.1 XP_008767885.1 4 - 5 511 68

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1300001832.rn.1

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Recommended Assay Rattus norvegicus 1700 Probe spans exons

          Panther Classification:

          Molecular Function -

          scaffold/adaptor protein

          Gene Ontology Categories:

          Function(s) Process(es)

          skeletal muscle contraction
          cellular hyperosmotic response
          actin filament binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P20726 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 671674 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 459249 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 604903 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 952209 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 103300 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 844793 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 250778 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 577569 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 522254 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 518861 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 962891 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 912780 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 725163 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 143431 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 15062 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P20726 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P20726 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P20726 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P561048 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P260174 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 100747 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10769551 - -
          Affymetrix Rat Gene 1.0 ST Array 10769550 - -
          Affymetrix Rat Gene 1.0 ST Array 10769552 - -
          Affymetrix Rat Gene 1.0 ST Array 10769554 - -
          Affymetrix Rat Gene 1.0 ST Array 10769549 - -
          Affymetrix Rat Gene 1.0 ST Array 10769548 - -
          Affymetrix Rat Gene 1.1 ST Array 10769547 - -
          Rat Genome 230 2.0 Array 1373903_at - -
          Rat Genome 230 2.0 Array 1373903_PM_at - -
          Affymetrix HT Rat Focus Array 1373903_PM_at - -
          Affymetrix HT Rat Focus Array 1373903_at - -
          Affymetrix Rat Clariom S Assay PSR1300012963.rn.1 TC1300001832.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1300012962.rn.1 TC1300001832.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1300012964.rn.1 TC1300001832.rn.1 RTA TC
          Rat Expression Set 230 1373903_PM_at - -
          Rat Expression Set 230 1373903_at - -
          Affymetrix Rat Gene 1.0 ST Array 10769547 - -
          Rat Genome U34 Set rc_AA925708_at - -
          Rat Genome U34 Set rc_AI011920_at - -
          Rat Genome U34 Set rc_AA901326_at - -
          Rat Genome U34 Set rc_AA925708_g_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21662423 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1359700 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline