Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01483492_m1
          See other SLC45A3 GE Assays ›
          Gene Symbol:
          Slc45a3
          Gene Name:
          solute carrier family 45, member 3
          Gene Aliases:
          RGD1309764
          Chromosome Location:
          Chr.13: 48745860 - 48766283 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01483492_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Slc45a3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001135868.1 NP_001129340.1 4 - 5 1565 59
          XM_006249765.2 XP_006249827.1 4 - 5 1540 59
          XM_006249766.2 XP_006249828.1 4 - 5 1665 59
          XM_006249767.2 XP_006249829.1 4 - 5 2143 59
          XM_006249769.2 XP_006249831.1 4 - 5 2315 59

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1300000248.rn.1

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Panther Classification:

          Molecular Function -

          secondary carrier transporter

          Gene Ontology Categories:

          Function(s)

          transmembrane transport

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P10089 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P022045 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 961688 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 3989 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 108968 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 893250 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 743722 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 678820 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 74263 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 968215 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 165988 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 273022 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 100004 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 547269 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 870999 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 531164 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 84640 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 12790 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P10089 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P022045 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P022045 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P10089 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P022045 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P10089 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P129298 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P10089 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10763867 - -
          Affymetrix Rat Gene 1.0 ST Array 10763865 - -
          Affymetrix Rat Gene 1.0 ST Array 10763868 - -
          Affymetrix Rat Gene 1.0 ST Array 10763866 - -
          Affymetrix Rat Gene 1.0 ST Array 10763864 - -
          Affymetrix Rat Gene 1.1 ST Array 10763863 - -
          Rat Genome 230 2.0 Array 1389113_at - -
          Rat Genome 230 2.0 Array 1389113_PM_at - -
          Affymetrix HT Rat Focus Array 1389113_at - -
          Affymetrix HT Rat Focus Array 1389113_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1300001464.rn.1 TC1300000248.rn.1 RTA TC
          Rat Expression Set 230 1389113_at - -
          Rat Expression Set 230 1389113_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10763863 - -
          Rat Genome U34 Set rc_AI137795_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21283941 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1371059 - -
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1530451 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline