Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01483520_m1
          See other RASAL2 GE Assays ›
          Gene Symbol:
          Rasal2
          Gene Name:
          RAS protein activator like 2
          Gene Aliases:
          -
          Chromosome Location:
          Chr.13: 74770557 - 74933964 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01483520_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rasal2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001107188.1 NP_001100658.1 5 - 6 588 68
          XM_006250065.2 XP_006250127.1 5 - 6 1078 68
          XM_006250066.2 XP_006250128.1 5 - 6 1687 68
          XM_006250067.2 XP_006250129.1 5 - 6 2546 68
          XM_008769635.1 XP_008767857.1 5 - 6 1078 68
          XM_008769636.1 XP_008767858.1 5 - 6 1078 68
          XM_008769637.1 XP_008767859.1 5 - 6 1078 68
          XM_008769638.1 XP_008767860.1 6 - 7 731 68
          XM_008769639.1 XP_008767861.1 6 - 7 731 68

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1300001706.rn.1

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Recommended Assay Rattus norvegicus 1700 Probe spans exons

          Panther Classification:

          Molecular Function -

          GTPase-activating protein

          Gene Ontology Categories:

          Function(s)

          signal transduction
          negative regulation of Ras protein signal transduction

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P321075 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 949732 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 719863 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 98322 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 160176 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 721745 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 71012 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 4907 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 490830 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 568111 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 757094 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 218812 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 235848 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 302135 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 183607 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 453751 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 327609 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 96854 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 357261 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 500037 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 123411 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 751967 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 296655 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 622340 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 519023 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 31525 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 511303 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 400607 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 270186 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 574868 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 550223 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 642219 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 617873 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 449768 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 930193 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 157472 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 21130 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 855941 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 554967 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 186672 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 488994 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 408103 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 115413 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 971000 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 502897 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 219180 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 961956 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 162848 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 764579 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 329480 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 652830 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 799696 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 662690 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 937880 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 527491 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 637683 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 95482 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 732516 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 58192 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 236317 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 863792 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 674154 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 969255 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 645593 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 68237 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 310140 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 963512 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 357133 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 335857 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 428253 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 583418 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 662410 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 298559 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 151276 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 470849 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 747556 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 913736 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 261566 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 605002 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 333011 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 427538 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 75786 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 400925 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 721773 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 739606 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 221021 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 488892 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 117484 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 141792 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 805451 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 175845 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 274022 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 782579 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 476898 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 531334 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 932765 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 87142 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 238657 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 141576 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 714316 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 612370 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 55895 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 112036 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 146659 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 93778 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 39405 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 94149 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 153860 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 128179 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 108112 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 97593 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 92782 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 140544 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 103633 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 145177 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 121121 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P321075 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P321075 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P321075 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P1051678 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P321075 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 78295 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 87320 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 33625 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 11925 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10769065 - -
          Affymetrix Rat Gene 1.0 ST Array 10769048 - -
          Affymetrix Rat Gene 1.0 ST Array 10769051 - -
          Affymetrix Rat Gene 1.0 ST Array 10769068 - -
          Affymetrix Rat Gene 1.0 ST Array 10769052 - -
          Affymetrix Rat Gene 1.0 ST Array 10769062 - -
          Affymetrix Rat Gene 1.0 ST Array 10769050 - -
          Affymetrix Rat Gene 1.0 ST Array 10769063 - -
          Affymetrix Rat Gene 1.0 ST Array 10769066 - -
          Affymetrix Rat Gene 1.0 ST Array 10769049 - -
          Affymetrix Rat Gene 1.0 ST Array 10769064 - -
          Affymetrix Rat Gene 1.0 ST Array 10769069 - -
          Affymetrix Rat Gene 1.0 ST Array 10769067 - -
          Affymetrix Rat Gene 1.1 ST Array 10769047 - -
          Rat Genome 230 2.0 Array 1378740_PM_at - -
          Rat Genome 230 2.0 Array 1378740_at - -
          Affymetrix HT Rat Focus Array 1378740_PM_at - -
          Affymetrix HT Rat Focus Array 1378740_at - -
          Affymetrix Rat Clariom S Assay PSR1300011957.rn.1 TC1300001706.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1300011959.rn.1 TC1300001706.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1300011963.rn.1 TC1300001706.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1300011964.rn.1 TC1300001706.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1300011961.rn.1 TC1300001706.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1300011958.rn.1 TC1300001706.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1300011967.rn.1 TC1300001706.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1300011965.rn.1 TC1300001706.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1300011973.rn.1 TC1300001706.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1300011974.rn.1 TC1300001706.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1300011962.rn.1 TC1300001706.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1300011955.rn.1 TC1300001706.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1300011975.rn.1 TC1300001706.rn.1 RTA TC
          Rat Expression Set 230 1378740_at - -
          Rat Expression Set 230 1378740_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10769047 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21244217 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1369925 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline