Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01483659_m1
          See other CDC73 GE Assays ›
          Gene Symbol:
          Cdc73
          Gene Name:
          cell division cycle 73
          Gene Aliases:
          Hrpt2
          Chromosome Location:
          Chr.13: 60403771 - 60496511 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01483659_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cdc73
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001024769.1 NP_001019940.1 15 - 16 1554 151
          GenBank mRNA BC097445.1 - 15 - 16 1554 7456471 151

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1300001548.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Rattus norvegicus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Cdc73

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          endodermal cell fate commitment
          transcription, DNA-templated
          mRNA polyadenylation
          biological_process
          negative regulation of cell proliferation
          histone monoubiquitination
          Wnt signaling pathway
          stem cell population maintenance
          positive regulation of Wnt signaling pathway
          positive regulation of mRNA 3'-end processing
          protein destabilization
          positive regulation of transcription elongation from RNA polymerase II promoter
          histone H2B ubiquitination
          negative regulation of myeloid cell differentiation
          positive regulation of transcription from RNA polymerase II promoter
          negative regulation of fibroblast proliferation
          negative regulation of epithelial cell proliferation
          cellular response to lipopolysaccharide
          negative regulation of G1/S transition of mitotic cell cycle
          RNA polymerase II core binding
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P22746 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 294638 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 403704 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 934887 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 408275 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 841611 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 916694 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 259154 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 37849 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 425973 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 921962 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 818997 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 803038 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 529377 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 21193 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 773164 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 932836 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 124970 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 263197 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 746478 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 906493 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 157094 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 125717 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 309256 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 874690 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 399952 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 284681 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 180207 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 930079 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 6830 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 790621 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 430216 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 191787 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 149471 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 844267 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 112012 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 84535 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 300395 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 729229 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 828278 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 17068 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 55368 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 816210 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 95633 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 552571 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 112567 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 546556 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 797518 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 580843 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 871649 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 365198 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 338242 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 741725 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 662604 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 758544 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 892745 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 525073 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 242980 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 266863 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 89830 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 483115 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 674342 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 748791 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 402124 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 101389 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 950857 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 198680 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 603888 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 26426 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 329588 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 238787 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 53803 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 124287 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 482780 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 52489 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 150889 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 61815 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 68029 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 132854 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 106960 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 71474 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 156558 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 102403 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P22746 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P22746 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P22746 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P22746 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P176494 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 62303 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 47026 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 103306 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 52231 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 62622 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 18016 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 53142 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 39676 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10768317 - -
          Affymetrix Rat Gene 1.0 ST Array 10768313 - -
          Affymetrix Rat Gene 1.0 ST Array 10768310 - -
          Affymetrix Rat Gene 1.0 ST Array 10768321 - -
          Affymetrix Rat Gene 1.0 ST Array 10768306 - -
          Affymetrix Rat Gene 1.0 ST Array 10768314 - -
          Affymetrix Rat Gene 1.0 ST Array 10768315 - -
          Affymetrix Rat Gene 1.0 ST Array 10768307 - -
          Affymetrix Rat Gene 1.0 ST Array 10768308 - -
          Affymetrix Rat Gene 1.0 ST Array 10768318 - -
          Affymetrix Rat Gene 1.0 ST Array 10768320 - -
          Affymetrix Rat Gene 1.0 ST Array 10768311 - -
          Affymetrix Rat Gene 1.0 ST Array 10768309 - -
          Affymetrix Rat Gene 1.0 ST Array 10768319 - -
          Affymetrix Rat Gene 1.0 ST Array 10768316 - -
          Affymetrix Rat Gene 1.0 ST Array 10768312 - -
          Affymetrix Rat Gene 1.0 ST Array 10768322 - -
          Affymetrix Rat Gene 1.1 ST Array 10768305 - -
          Rat Genome 230 2.0 Array 1379346_at - -
          Rat Genome 230 2.0 Array 1379346_PM_at - -
          Affymetrix HT Rat Focus Array 1379346_PM_at - -
          Affymetrix HT Rat Focus Array 1379346_at - -
          Affymetrix Rat Clariom S Assay PSR1300010637.rn.1 TC1300001548.rn.1 RTA TC
          Rat Expression Set 230 1379346_PM_at - -
          Rat Expression Set 230 1379346_at - -
          Affymetrix Rat Gene 1.0 ST Array 10768305 - -
          Rat Genome U34 Set rc_AI013310_at - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1358656 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline