Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01483765_m1
          See other NUF2 GE Assays ›
          Gene Symbol:
          Nuf2
          Gene Name:
          NUF2, NDC80 kinetochore complex component
          Gene Aliases:
          Cdca1
          Chromosome Location:
          Chr.13: 87818456 - 87847223 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01483765_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Nuf2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001012028.1 NP_001012028.1 2 - 3 257 111
          GenBank mRNA BC078993.1 - 2 - 3 257 7112621 111

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1300001870.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Nuf2

          Panther Classification:

          Molecular Function -

          non-motor microtubule binding protein

          Gene Ontology Categories:

          Function(s)

          mitotic nuclear division
          cell division

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P1005213 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 215702 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 517985 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 731669 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 614458 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 329655 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 589169 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 224190 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 682434 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 622846 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 866241 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 945454 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 345313 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 962028 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 97539 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 396097 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 122577 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 712524 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 47300 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 609170 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 157994 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 57918 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 549823 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 3448 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 117886 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 141291 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P1005213 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P1005213 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P1005213 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P1005213 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 63074 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 45007 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 65599 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10769669 - -
          Affymetrix Rat Gene 1.0 ST Array 10769671 - -
          Affymetrix Rat Gene 1.0 ST Array 10769662 - -
          Affymetrix Rat Gene 1.0 ST Array 10769661 - -
          Affymetrix Rat Gene 1.0 ST Array 10769667 - -
          Affymetrix Rat Gene 1.0 ST Array 10769666 - -
          Affymetrix Rat Gene 1.0 ST Array 10769665 - -
          Affymetrix Rat Gene 1.0 ST Array 10769663 - -
          Affymetrix Rat Gene 1.0 ST Array 10769668 - -
          Affymetrix Rat Gene 1.0 ST Array 10769670 - -
          Affymetrix Rat Gene 1.0 ST Array 10769660 - -
          Affymetrix Rat Gene 1.0 ST Array 10769659 - -
          Affymetrix Rat Gene 1.0 ST Array 10769664 - -
          Affymetrix Rat Gene 1.1 ST Array 10769657 - -
          Rat Genome 230 2.0 Array 1383940_PM_at - -
          Rat Genome 230 2.0 Array 1383940_at - -
          Affymetrix HT Rat Focus Array 1383940_at - -
          Affymetrix HT Rat Focus Array 1383940_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1300013195.rn.1 TC1300001870.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1300013200.rn.1 TC1300001870.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1300013203.rn.1 TC1300001870.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1300013204.rn.1 TC1300001870.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1300013207.rn.1 TC1300001870.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1300013196.rn.1 TC1300001870.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1300013201.rn.1 TC1300001870.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1300013209.rn.1 TC1300001870.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1300013199.rn.1 TC1300001870.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1300013208.rn.1 TC1300001870.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1300013197.rn.1 TC1300001870.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1300013206.rn.1 TC1300001870.rn.1 RTA TC
          Rat Expression Set 230 1383940_PM_at - -
          Rat Expression Set 230 1383940_at - -
          Affymetrix Rat Gene 1.0 ST Array 10769657 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21917376 - -
          ABI Rat Genome Survey Microarray 22041071 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1375974 - -
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1376705 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline